This product has been added to your favorites list. Go to My Favorites

System Message

OKCancel
LOADING ...
  • Home
  • › Search Tool
  • › Search Results
  • › At02186798_g1
See other PDIL1-4 GE Assays ›
Gene Symbol:
PDIL1-4
Gene Name:
PDI-like 1-4
Gene Aliases:
ARABIDOPSIS THALIANA PROTEIN DISULFIDE ISOMERASE 2, AT5G60640, ATPDI2, ATPDIL1-4, MUP24.6, MUP24_6, PDI-like 1-4, PDI2, PROTEIN DISULFIDE ISOMERASE 2
Chromosome Location:
Chr.5: 24370827 - 24374098 on Build TAIR10.1
Species:
Arabidopsis
Species Specific ID (Flybase ID):
-
Assay ID At02186798_g1
Size
Availability Made To Order
Catalog # 4351372
Price
Your Price
Online offer:
Check your price ›
  • Genomic Map
  • Assay Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

Assay Details

Gene Symbol: PDIL1-4
Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
RefSeq NM_001203652.1 NP_001190581.1 4 - 5 1046 109
NM_125457.3 NP_568926.2 4 - 5 1046 109
NM_180903.4 NP_851234.1 4 - 5 1106 109
GenBank mRNA BT001994.1 - 4 - 5 1044 109
BT008359.1 - 4 - 5 1001 109

Back To Top

More Information


Set Membership:

Amplicon length greater than or equal to 101 Detect Genomic DNA Arabidopsis thaliana Probe spans exons

Panther Classification:

Molecular Function -

chaperone

Gene Ontology Categories:

Function(s) Process(es)

protein folding
response to oxidative stress
response to endoplasmic reticulum stress
cell redox homeostasis
protein disulfide isomerase activity

Back To Top

Related Products

  • TaqMan® Fast Advanced Master Mix
  • SuperScript® VILO™ cDNA Synthesis Kit

Your items have has been added!


Host server : magellan-search-5fc94fc45-hqqk4:80/100.66.128.142:80.
git-commit: 7a6627b71d15189168043ee5c63248cc100e5965
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.37.0-2025.10.41-1.0