This product has been added to your favorites list. Go to My Favorites

System Message

OKCancel
LOADING ...
  • Home
  • › Search Tool
  • › Search Results
  • › At02186517_g1
See other RPL24 GE Assays ›
Gene Symbol:
RPL24
Gene Name:
Translation protein SH3-like family protein
Gene Aliases:
AT5G54600, MRB17.10, MRB17_10, SUPPRESSOR OF VARIEGATION 8, SVR8, plastid ribosomal protein L24
Chromosome Location:
Chr.5: 22182843 - 22184610 on Build TAIR10.1
Species:
Arabidopsis
Species Specific ID (Flybase ID):
-
Assay ID At02186517_g1
Best Coverage
Size
Availability Made To Order
Catalog # 4351372
Price 374.00
Your Price
Online offer:
374.0
Check your price ›
  • Genomic Map
  • Assay Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

Assay Details

Gene Symbol: RPL24
Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
RefSeq NM_124841.2 NP_200271.2 2 - 3 222 70
NM_180859.2 NP_851190.1 2 - 3 357 70
GenBank mRNA AY086810.1 - 2 - 3 222 70
BT001990.1 - 2 - 3 191 70
BT006573.1 - 2 - 3 154 70

Back To Top

More Information


Set Membership:

Amplicon length less than or equal to 70 3' Most Detect Genomic DNA Recommended Assay Arabidopsis thaliana Probe spans exons

Panther Classification:

Molecular Function -

ribosomal protein

Gene Ontology Categories:

Function(s) Process(es)

translation
plastid translation
RNA binding
mRNA binding
structural constituent of ribosome

Back To Top

Related Products

  • TaqMan® Fast Advanced Master Mix
  • SuperScript® VILO™ cDNA Synthesis Kit

Your items have has been added!


Host server : magellan-search-5fc94fc45-hqqk4:80/100.66.128.142:80.
git-commit: 7a6627b71d15189168043ee5c63248cc100e5965
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.37.0-2025.10.41-1.0