This product has been added to your favorites list. Go to My Favorites

System Message

OKCancel
LOADING ...
  • Home
  • › Search Tool
  • › Search Results
  • › At02185462_g1
See other AT3G63460 GE Assays ›
Gene Symbol:
AT3G63460
Gene Name:
transducin family protein / WD-40 repeat family protein
Gene Aliases:
AT3G63460, SEC31B
Chromosome Location:
Chr.3: 23430623 - 23437374 on Build TAIR10.1
Species:
Arabidopsis
Species Specific ID (Flybase ID):
-
Assay ID At02185462_g1
Size
Availability Made To Order
Catalog # 4351372
Price
Your Price
Online offer:
Check your price ›
  • Genomic Map
  • Assay Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

Assay Details

Gene Symbol: AT3G63460
Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
RefSeq NM_001203242.1 NP_001190171.1 16 - 17 2459 90
NM_116211.3 NP_191905.3 16 - 17 2453 90
NM_180693.2 NP_851024.1 16 - 17 2482 90
GenBank mRNA AY099702.1 - 16 - 17 2423 90
BT008435.1 - 16 - 17 2356 90

Back To Top

More Information


Set Membership:

Amplicon length between 86 and 100 Detect Genomic DNA Arabidopsis thaliana Probe spans exons

Panther Classification:

Molecular Function -

vesicle coat protein

Gene Ontology Categories:

Function(s) Process(es)

intracellular protein transport
ER to Golgi vesicle-mediated transport
endoplasmic reticulum organization
cargo loading into COPII-coated vesicle
COPII-coated vesicle budding
structural molecule activity

Back To Top

Related Products

  • TaqMan® Fast Advanced Master Mix
  • SuperScript® VILO™ cDNA Synthesis Kit

Your items have has been added!


Host server : magellan-search-56696975f8-skfwn:80/100.66.128.142:80.
git-commit: 795bdb1f794b4b81290d0c522fee4b91276a707f
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.36.2-2025.09.39-1.0