This product has been added to your favorites list. Go to My Favorites

System Message

OKCancel
LOADING ...
  • Home
  • › Search Tool
  • › Search Results
  • › At02185141_g1
See other ALDH11A3 GE Assays ›
Gene Symbol:
ALDH11A3
Gene Name:
aldehyde dehydrogenase 11A3
Gene Aliases:
AT2G24270, F27D4.18, F27D4_18, NP-GAPDH, aldehyde dehydrogenase 11A3
Chromosome Location:
Chr.2: 10326918 - 10330048 on Build TAIR10.1
Species:
Arabidopsis
Species Specific ID (Flybase ID):
-
Assay ID At02185141_g1
Size
Availability Made To Order
Catalog # 4351372
Price 374.00
Your Price
Online offer:
374.0
Check your price ›
  • Genomic Map
  • Assay Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

Assay Details

Gene Symbol: ALDH11A3
Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
RefSeq NM_001202660.1 NP_001189589.1 7 - 8 1108 116
NM_001202661.1 NP_001189590.1 7 - 8 1119 116
NM_001335927.1 NP_001318281.1 7 - 8 1186 116
NM_127989.4 NP_180004.1 6 - 7 1329 116
GenBank mRNA AK317484.1 - 7 - 8 947 116
AY037205.1 - 6 - 7 1000 116
AY136409.1 - 6 - 7 1085 116
BT004551.1 - 6 - 7 882 116

Back To Top

More Information


Set Membership:

Amplicon length greater than or equal to 101 Detect Genomic DNA Arabidopsis thaliana Probe spans exons

Panther Classification:

Molecular Function -

dehydrogenase

Gene Ontology Categories:

Function(s) Process(es)

oxidation-reduction process
glyceraldehyde-3-phosphate dehydrogenase (NADP+) (non-phosphorylating) activity

Back To Top

Related Products

  • TaqMan® Fast Advanced Master Mix
  • SuperScript® VILO™ cDNA Synthesis Kit

Your items have has been added!


Host server : magellan-search-5fc94fc45-hqqk4:80/100.66.128.142:80.
git-commit: 7a6627b71d15189168043ee5c63248cc100e5965
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.37.0-2025.10.41-1.0