This product has been added to your favorites list. Go to My Favorites

System Message

OKCancel
LOADING ...
  • Home
  • › Search Tool
  • › Search Results
  • › At02184873_g1
See other SUVR5 GE Assays ›
Gene Symbol:
SUVR5
Gene Name:
histone-lysine N-methyltransferase SUVR5
Gene Aliases:
AT2G23740, F27L4.8, F27L4_8, SU(VAR)3-9-RELATED protein 5
Chromosome Location:
Chr.2: 10097265 - 10103398 on Build TAIR10.1
Species:
Arabidopsis
Species Specific ID (Flybase ID):
-
Assay ID At02184873_g1
Best Coverage
Size
Availability Made To Order
Catalog # 4351372
Price 374.00
Your Price
Online offer:
374.0
Check your price ›
  • Genomic Map
  • Assay Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

Assay Details

Gene Symbol: SUVR5
Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
RefSeq NM_001202656.2 NP_001189585.1 2 - 3 600 72
NM_001335861.1 NP_001324207.1 2 - 3 481 72
NM_001335862.1 NP_001324206.1 2 - 3 341 72
NM_001335863.1 NP_001324205.1 2 - 3 569 72
NM_001335864.1 NP_001324204.1 2 - 3 382 72
NM_127937.2 NP_179954.2 2 - 3 520 72
GenBank mRNA AK318822.1 - 2 - 3 520 72

Back To Top

More Information


Set Membership:

Amplicon length between 71 and 85 Detect Genomic DNA Recommended Assay Arabidopsis thaliana Probe spans exons

Panther Classification:

Molecular Function -

histone modifying enzyme

Gene Ontology Categories:

Function(s) Process(es)

chromatin remodeling
chromatin silencing
histone lysine methylation
regulation of histone H3-K9 dimethylation
transcription factor activity, sequence-specific DNA binding
zinc ion binding
histone-lysine N-methyltransferase activity
sequence-specific DNA binding

Back To Top

Related Products

  • TaqMan® Fast Advanced Master Mix
  • SuperScript® VILO™ cDNA Synthesis Kit

Your items have has been added!


Host server : magellan-search-69fc894774-tvlwx:80/100.66.128.142:80.
git-commit: 8e82defa9bf25b3b77800b5e306013d4ba941391
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.36.2-2025.09.39-1.0