This product has been added to your favorites list. Go to My Favorites

System Message

OKCancel
LOADING ...
  • Home
  • › Search Tool
  • › Search Results
  • › At02184306_m1
See other AGL17 GE Assays ›
Gene Symbol:
AGL17
Gene Name:
AGAMOUS-like 17
Gene Aliases:
AGAMOUS-like 17, AT2G22630, T9I22.7, T9I22_7
Chromosome Location:
Chr.2: 9618207 - 9622163 on Build TAIR10.1
Species:
Arabidopsis
Species Specific ID (Flybase ID):
-
Assay ID At02184306_m1
Best Coverage
Size
Availability Made To Order
Catalog # 4351372
Price 374.00
Your Price
Online offer:
374.0
Check your price ›
  • Genomic Map
  • Assay Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

Assay Details

Gene Symbol: AGL17
Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
RefSeq NM_001335805.1 NP_001324454.1 1 - 2 349 76
NM_127828.3 NP_179848.1 1 - 2 349 76
GenBank mRNA AB493558.1 - 1 - 2 184 76

Back To Top

More Information


Set Membership:

Amplicon length between 71 and 85 Recommended Assay Arabidopsis thaliana Probe spans exons

Gene Ontology Categories:

Function(s) Process(es)

regulation of transcription, DNA-templated
positive regulation of transcription from RNA polymerase II promoter
positive regulation of long-day photoperiodism, flowering
RNA polymerase II regulatory region sequence-specific DNA binding
transcription factor activity, sequence-specific DNA binding
protein binding
protein dimerization activity

Back To Top

Related Products

  • TaqMan® Fast Advanced Master Mix
  • SuperScript® VILO™ cDNA Synthesis Kit

Your items have has been added!


Host server : magellan-search-5c796d7db4-nk9d7:80/100.66.128.142:80.
git-commit: d3b3aa98da0ae1b146c174fddb0766cfa6979daf
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.36.2-2025.09.39-1.0