This product has been added to your favorites list. Go to My Favorites

System Message

OKCancel
LOADING ...
  • Home
  • › Search Tool
  • › Search Results
  • › At02184217_m1
See other SVP GE Assays ›
Gene Symbol:
SVP
Gene Name:
K-box region and MADS-box transcription factor family protein
Gene Aliases:
AGAMOUS-like 22, AGL22, AT2G22540, AT2G22550, F14M13.6, F14M13_6, FAQ1, Flowering Arabidopsis QTL1, SHORT VEGETATIVE PHASE
Chromosome Location:
Chr.2: 9579647 - 9583901 on Build TAIR10.1
Species:
Arabidopsis
Species Specific ID (Flybase ID):
-
Assay ID At02184217_m1
Size
Availability Made To Order
Catalog # 4351372
Price
Your Price
Online offer:
Check your price ›
  • Genomic Map
  • Assay Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

Assay Details

Gene Symbol: SVP
Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
RefSeq NM_001335800.1 NP_001324584.1 2 - 3 291 90
NM_127820.4 NP_179840.2 1 - 2 954 90
GenBank mRNA AF211171.1 - 1 - 2 713 90
BT033098.1 - 1 - 2 184 90
EU078687.1 - 1 - 2 184 90

Back To Top

More Information


Set Membership:

Amplicon length between 86 and 100 Arabidopsis thaliana Probe spans exons

Gene Ontology Categories:

Function(s) Process(es)

response to temperature stimulus
negative regulation of flower development
maintenance of floral meristem identity
floral meristem determinacy
negative regulation of transcription, DNA-templated
positive regulation of transcription from RNA polymerase II promoter
floral whorl development
translation repressor activity, nucleic acid binding
transcription regulatory region sequence-specific DNA binding
RNA polymerase II regulatory region sequence-specific DNA binding
RNA polymerase II transcription factor activity, sequence-specific DNA binding
transcription factor activity, sequence-specific DNA binding
protein binding
transcription factor binding
sequence-specific DNA binding
protein dimerization activity

Back To Top

Related Products

  • TaqMan® Fast Advanced Master Mix
  • SuperScript® VILO™ cDNA Synthesis Kit

Your items have has been added!


Host server : magellan-search-5fc94fc45-hqqk4:80/100.66.128.142:80.
git-commit: 7a6627b71d15189168043ee5c63248cc100e5965
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.37.0-2025.10.41-1.0