This product has been added to your favorites list. Go to My Favorites

System Message

OKCancel
LOADING ...
  • Home
  • › Search Tool
  • › Search Results
  • › At02184175_s1
See other UCP5 GE Assays ›
Gene Symbol:
UCP5
Gene Name:
uncoupling protein 5
Gene Aliases:
AT2G22500, ATPUMP5, DIC1, DICARBOXYLATE CARRIER 1, F14M13.10, F14M13_10, PLANT UNCOUPLING MITOCHONDRIAL PROTEIN 5, uncoupling protein 5
Chromosome Location:
Chr.2: 9562774 - 9564850 on Build TAIR10.1
Species:
Arabidopsis
Species Specific ID (Flybase ID):
-
Assay ID At02184175_s1
Best Coverage
Size
Availability Made To Order
Catalog # 4351372
Price
Your Price
Online offer:
Check your price ›
  • Genomic Map
  • Assay Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

Assay Details

Gene Symbol: UCP5
Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
RefSeq NM_127816.3 NP_179836.1 1 1164 70
GenBank mRNA AF370588.1 - 1 786 70
AM236862.1 - 1 1050 70
AY142648.1 - 1 786 70

Back To Top

More Information


Set Membership:

Amplicon length less than or equal to 70 3' Most Detect Genomic DNA Recommended Assay Arabidopsis thaliana Both primers and probe map within a single exon

Panther Classification:

Molecular Function -

transporter secondary carrier transporter

Gene Ontology Categories:

Function(s) Process(es)

mitochondrial transport
sulfate transport
thiosulfate transport
oxaloacetate transport
phosphate ion transmembrane transport
succinate transmembrane transport
malate transmembrane transport
cellular response to hypoxia
hydrogen ion transmembrane transport
dicarboxylic acid transmembrane transporter activity
sulfate transmembrane transporter activity
thiosulfate transmembrane transporter activity
oxaloacetate transmembrane transporter activity
malate transmembrane transporter activity
succinate transmembrane transporter activity
antiporter activity
oxidative phosphorylation uncoupler activity

Back To Top

Related Products

  • TaqMan® Fast Advanced Master Mix
  • SuperScript® VILO™ cDNA Synthesis Kit

Your items have has been added!


Host server : magellan-search-5fc94fc45-hqqk4:80/100.66.128.142:80.
git-commit: 7a6627b71d15189168043ee5c63248cc100e5965
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.37.0-2025.10.41-1.0