This product has been added to your favorites list. Go to My Favorites

System Message

OKCancel
LOADING ...
  • Home
  • › Search Tool
  • › Search Results
  • › At02183042_g1
See other SMC6A GE Assays ›
Gene Symbol:
SMC6A
Gene Name:
structural maintenance of chromosomes 6A
Gene Aliases:
AT5G07660, MBK20.10, MBK20_10, structural maintenance of chromosomes 6A
Chromosome Location:
Chr.5: 2422783 - 2430136 on Build TAIR10.1
Species:
Arabidopsis
Species Specific ID (Flybase ID):
-
Assay ID At02183042_g1
Size
Availability Made To Order
Catalog # 4351372
Price
Your Price
Online offer:
Check your price ›
  • Genomic Map
  • Assay Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

Assay Details

Gene Symbol: SMC6A
Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
RefSeq NM_120848.2 NP_196383.1 17 - 18 2223 129

Back To Top

More Information


Set Membership:

Amplicon length greater than or equal to 101 Detect Genomic DNA Arabidopsis thaliana Probe spans exons

Panther Classification:

Molecular Function -

DNA metabolism protein

Gene Ontology Categories:

Function(s)

double-strand break repair via homologous recombination
sister chromatid cohesion
response to X-ray

Back To Top

Related Products

  • TaqMan® Fast Advanced Master Mix
  • SuperScript® VILO™ cDNA Synthesis Kit

Your items have has been added!


Host server : magellan-search-6f7678f454-w8g9k:80/100.66.128.142:80.
git-commit: 7b0558793d0e70b0085ba95ba375cdfe261f8cbf
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.36.2-2025.09.39-1.0