This product has been added to your favorites list. Go to My Favorites

System Message

OKCancel
LOADING ...
  • Home
  • › Search Tool
  • › Search Results
  • › At02181608_g1
See other EIF2 ALPHA GE Assays ›
Gene Symbol:
EIF2 ALPHA
Gene Name:
eukaryotic translation initiation factor 2 alpha subunit
Gene Aliases:
AT5G05470, ATEIF2-A2, EIF2-A2, eukaryotic translation initiation factor 2 alpha subunit
Chromosome Location:
Chr.5: 1620458 - 1622340 on Build TAIR10.1
Species:
Arabidopsis
Species Specific ID (Flybase ID):
-
Assay ID At02181608_g1
Size
Availability Made To Order
Catalog # 4351372
Price 374.00
Your Price
Online offer:
374.0
Check your price ›
  • Genomic Map
  • Assay Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

Assay Details

Gene Symbol: EIF2 ALPHA
Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
RefSeq NM_120629.3 NP_196166.1 6 - 7 996 85
GenBank mRNA AF324988.2 - 6 - 7 996 85
AK117513.1 - 6 - 7 990 85
BT024879.1 - 6 - 7 875 85

Back To Top

More Information


Set Membership:

Amplicon length between 71 and 85 3' Most Detect Genomic DNA Arabidopsis thaliana Probe spans exons

Panther Classification:

Molecular Function -

translation initiation factor

Gene Ontology Categories:

Process(es)

RNA binding
translation initiation factor activity
ribosome binding

Back To Top

Related Products

  • TaqMan® Fast Advanced Master Mix
  • SuperScript® VILO™ cDNA Synthesis Kit

Your items have has been added!


Host server : magellan-search-5fc94fc45-4mbwj:80/100.66.130.94:80.
git-commit: 7a6627b71d15189168043ee5c63248cc100e5965
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.37.0-2025.10.41-1.0