This product has been added to your favorites list. Go to My Favorites

System Message

OKCancel
LOADING ...
  • Home
  • › Search Tool
  • › Search Results
  • › At02180995_s1
See other SIM GE Assays ›
Gene Symbol:
SIM
Gene Name:
cyclin-dependent protein kinase inhibitor
Gene Aliases:
AT5G04470, SIAMESE, T19N18.8
Chromosome Location:
Chr.5: 1266625 - 1267416 on Build TAIR10.1
Species:
Arabidopsis
Species Specific ID (Flybase ID):
-
Assay ID At02180995_s1
Best Coverage
Size
Availability Made To Order
Catalog # 4351372
Price
Your Price
Online offer:
Check your price ›
  • Genomic Map
  • Assay Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

Assay Details

Gene Symbol: SIM
Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
RefSeq NM_120529.3 NP_196067.1 1 684 80
GenBank mRNA AY085204.1 - 1 637 80

Back To Top

More Information


Set Membership:

Amplicon length between 71 and 85 3' Most Detect Genomic DNA Recommended Assay Arabidopsis thaliana Both primers and probe map within a single exon

Panther Classification:

Molecular Function -

kinase inhibitor

Gene Ontology Categories:

Function(s) Process(es)

trichome differentiation
regulation of DNA endoreduplication
DNA endoreduplication
negative regulation of cyclin-dependent protein serine/threonine kinase activity
negative regulation of cell cycle
negative regulation of mitotic nuclear division
cyclin-dependent protein serine/threonine kinase inhibitor activity
protein binding

Back To Top

Related Products

  • TaqMan® Fast Advanced Master Mix
  • SuperScript® VILO™ cDNA Synthesis Kit

Your items have has been added!


Host server : magellan-search-5fc94fc45-hqqk4:80/100.66.128.142:80.
git-commit: 7a6627b71d15189168043ee5c63248cc100e5965
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.37.0-2025.10.41-1.0