This product has been added to your favorites list. Go to My Favorites

System Message

OKCancel
LOADING ...
  • Home
  • › Search Tool
  • › Search Results
  • › At02178734_g1
See other TIM GE Assays ›
Gene Symbol:
TIM
Gene Name:
triosephosphate isomerase
Gene Aliases:
AT2G21170, F26H11.7, F26H11_7, PDTPI, PLASTID ISOFORM TRIOSE PHOSPHATE ISOMERASE, TRIOSEPHOSPHATE ISOMERASE, triosephosphate isomerase
Chromosome Location:
Chr.2: 9070734 - 9074470 on Build TAIR10.1
Species:
Arabidopsis
Species Specific ID (Flybase ID):
-
Assay ID At02178734_g1
Size
Availability Made To Order
Catalog # 4351372
Price 374.00
Your Price
Online offer:
374.0
Check your price ›
  • Genomic Map
  • Assay Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

Assay Details

Gene Symbol: TIM
Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
RefSeq NM_001084462.1 NP_001077931.1 7 - 8 993 98
NM_001335732.1 NP_001323967.1 9 - 10 1155 98
NM_127687.4 NP_179713.1 8 - 9 1116 98
GenBank mRNA AF247559.1 - 8 - 9 953 98
AF378898.1 - 8 - 9 942 98
AY052748.1 - 8 - 9 893 98
AY087893.1 - 8 - 9 987 98

Back To Top

More Information


Set Membership:

Amplicon length between 86 and 100 3' Most Detect Genomic DNA Arabidopsis thaliana Probe spans exons

Panther Classification:

Molecular Function -

isomerase

Gene Ontology Categories:

Function(s) Process(es)

gluconeogenesis
glycolytic process
triglyceride mobilization
chloroplast organization
reductive pentose-phosphate cycle
glycerol catabolic process
multicellular organism reproduction
glyceraldehyde-3-phosphate biosynthetic process
primary root development
triose-phosphate isomerase activity

Back To Top

Related Products

  • TaqMan® Fast Advanced Master Mix
  • SuperScript® VILO™ cDNA Synthesis Kit

Your items have has been added!


Host server : magellan-search-5fc94fc45-4mbwj:80/100.66.130.94:80.
git-commit: 7a6627b71d15189168043ee5c63248cc100e5965
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.37.0-2025.10.41-1.0