This product has been added to your favorites list. Go to My Favorites

System Message

OKCancel
LOADING ...
  • Home
  • › Search Tool
  • › Search Results
  • › At02178304_g1
See other AT2G20420 GE Assays ›
Gene Symbol:
AT2G20420
Gene Name:
ATP citrate lyase (ACL) family protein
Gene Aliases:
AT2G20420, F11A3.3, F11A3_3
Chromosome Location:
Chr.2: 8805342 - 8808171 on Build TAIR10.1
Species:
Arabidopsis
Species Specific ID (Flybase ID):
-
Assay ID At02178304_g1
Size
Availability Made To Order
Catalog # 4351372
Price 374.00
Your Price
Online offer:
374.0
Check your price ›
  • Genomic Map
  • Assay Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

Assay Details

Gene Symbol: AT2G20420
Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
RefSeq NM_127601.4 NP_179632.1 1 - 2 327 77
GenBank mRNA AJ001808.1 - 1 - 2 179 77
AY099707.1 - 1 - 2 179 77
AY128889.1 - 1 - 2 95 77

Back To Top

More Information


Set Membership:

Amplicon length between 71 and 85 Detect Genomic DNA Arabidopsis thaliana Probe spans exons

Panther Classification:

Molecular Function -

ligase

Gene Ontology Categories:

Function(s) Process(es)

tricarboxylic acid cycle
succinyl-CoA metabolic process
response to cadmium ion
succinate-CoA ligase (ADP-forming) activity
succinate-CoA ligase (GDP-forming) activity
copper ion binding
ATP binding

Back To Top

Related Products

  • TaqMan® Fast Advanced Master Mix
  • SuperScript® VILO™ cDNA Synthesis Kit

Your items have has been added!


Host server : magellan-search-5c796d7db4-nk9d7:80/100.66.128.142:80.
git-commit: d3b3aa98da0ae1b146c174fddb0766cfa6979daf
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.36.2-2025.09.39-1.0