This product has been added to your favorites list. Go to My Favorites

System Message

OKCancel
LOADING ...
  • Home
  • › Search Tool
  • › Search Results
  • › At02177623_g1
See other AT2G19400 GE Assays ›
Gene Symbol:
AT2G19400
Gene Name:
AGC (cAMP-dependent, cGMP-dependent and protein kinase C) kinase family protein
Gene Aliases:
AT2G19400, F27F23.20, F27F23_20
Chromosome Location:
Chr.2: 8399100 - 8402664 on Build TAIR10.1
Species:
Arabidopsis
Species Specific ID (Flybase ID):
-
Assay ID At02177623_g1
Size
Availability Made To Order
Catalog # 4351372
Price
Your Price
Online offer:
Check your price ›
  • Genomic Map
  • Assay Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

Assay Details

Gene Symbol: AT2G19400
Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
RefSeq NM_127498.3 NP_565453.1 10 - 11 1576 98

Back To Top

More Information


Set Membership:

Amplicon length between 86 and 100 Detect Genomic DNA Arabidopsis thaliana Probe spans exons

Panther Classification:

Molecular Function -

non-receptor serine/threonine protein kinase

Gene Ontology Categories:

Function(s) Process(es)

phosphorylation
peptidyl-serine phosphorylation
intracellular signal transduction
protein serine/threonine kinase activity
kinase activity

Back To Top

Related Products

  • TaqMan® Fast Advanced Master Mix
  • SuperScript® VILO™ cDNA Synthesis Kit

Your items have has been added!


Host server : magellan-search-5fc94fc45-hqqk4:80/100.66.128.142:80.
git-commit: 7a6627b71d15189168043ee5c63248cc100e5965
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.37.0-2025.10.41-1.0