This product has been added to your favorites list. Go to My Favorites

System Message

OKCancel
LOADING ...
  • Home
  • › Search Tool
  • › Search Results
  • › At02176911_g1
See other KDTA GE Assays ›
Gene Symbol:
KDTA
Gene Name:
KDO transferase A
Gene Aliases:
AT5G03770, AtKdtA, F17C15.190, F17C15_190, KDO transferase A
Chromosome Location:
Chr.5: 995095 - 997849 on Build TAIR10.1
Species:
Arabidopsis
Species Specific ID (Flybase ID):
-
Assay ID At02176911_g1
Size
Availability Made To Order
Catalog # 4351372
Price
Your Price
Online offer:
Check your price ›
  • Genomic Map
  • Assay Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

Assay Details

Gene Symbol: KDTA
Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
RefSeq NM_001342729.1 NP_001331079.1 5 - 6 668 84
NM_001342730.1 NP_001331078.1 4 - 5 777 84
NM_120458.3 NP_195997.2 5 - 6 668 84
GenBank mRNA AY065115.1 - 5 - 6 603 84
AY128758.1 - 5 - 6 572 84
KJ138708.1 - 5 - 6 572 84

Back To Top

More Information


Set Membership:

Amplicon length between 71 and 85 Detect Genomic DNA Arabidopsis thaliana Probe spans exons

Panther Classification:

Molecular Function -

transferase

Gene Ontology Categories:

Function(s) Process(es)

lipid A biosynthetic process
Kdo2-lipid A biosynthetic process
transferase activity
transferase activity, transferring glycosyl groups
Kdo transferase activity

Back To Top

Related Products

  • TaqMan® Fast Advanced Master Mix
  • SuperScript® VILO™ cDNA Synthesis Kit

Your items have has been added!


Host server : magellan-search-7cdd86b58d-kqgnb:80/100.66.128.142:80.
git-commit: 00e1f9c870be1646ae16b59a7cca7819e45b49ab
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.36.2-2025.09.39-1.0