This product has been added to your favorites list. Go to My Favorites

System Message

OKCancel
LOADING ...
  • Home
  • › Search Tool
  • › Search Results
  • › At02176251_g1
See other GSTL3 GE Assays ›
Gene Symbol:
GSTL3
Gene Name:
Glutathione S-transferase family protein
Gene Aliases:
AT5G02790, F9G14.100, F9G14_100, Glutathione transferase L3
Chromosome Location:
Chr.5: 632796 - 635240 on Build TAIR10.1
Species:
Arabidopsis
Species Specific ID (Flybase ID):
-
Assay ID At02176251_g1
Size
Availability Made To Order
Catalog # 4351372
Price
Your Price
Online offer:
Check your price ›
  • Genomic Map
  • Assay Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

Assay Details

Gene Symbol: GSTL3
Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
RefSeq NM_120357.6 NP_195899.1 7 - 8 597 66
GenBank mRNA AK227162.1 - 7 - 8 552 66
AY140069.1 - 7 - 8 533 66
BT002166.1 - 7 - 8 516 66

Back To Top

More Information


Set Membership:

Amplicon length less than or equal to 70 Detect Genomic DNA Arabidopsis thaliana Probe spans exons

Panther Classification:

Molecular Function -

transferase metabolite interconversion enzyme

Gene Ontology Categories:

Function(s) Process(es)

protein glutathionylation
response to cadmium ion
glutathione transferase activity

Back To Top

Related Products

  • TaqMan® Fast Advanced Master Mix
  • SuperScript® VILO™ cDNA Synthesis Kit

Your items have has been added!


Host server : magellan-search-5c796d7db4-nk9d7:80/100.66.128.142:80.
git-commit: d3b3aa98da0ae1b146c174fddb0766cfa6979daf
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.36.2-2025.09.39-1.0