This product has been added to your favorites list. Go to My Favorites

System Message

OKCancel
LOADING ...
  • Home
  • › Search Tool
  • › Search Results
  • › At02176073_g1
See other ALG10 GE Assays ›
Gene Symbol:
ALG10
Gene Name:
DIE2/ALG10 family
Gene Aliases:
AT5G02410, T1E22.170, T1E22_170, homolog of yeast ALG10
Chromosome Location:
Chr.5: 516985 - 519850 on Build TAIR10.1
Species:
Arabidopsis
Species Specific ID (Flybase ID):
-
Assay ID At02176073_g1
Size
Availability Made To Order
Catalog # 4351372
Price
Your Price
Online offer:
Check your price ›
  • Genomic Map
  • Assay Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

Assay Details

Gene Symbol: ALG10
Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
RefSeq NM_120319.4 NP_195861.2 6 - 7 1343 149
GenBank mRNA AY099540.1 - 6 - 7 1249 149
BT002117.1 - 6 - 7 1127 149
KJ138711.1 - 6 - 7 1127 149

Back To Top

More Information


Set Membership:

Amplicon length greater than or equal to 101 Detect Genomic DNA Arabidopsis thaliana Probe spans exons

Panther Classification:

Molecular Function -

transferase

Gene Ontology Categories:

Function(s) Process(es)

protein N-linked glycosylation
dolichol-linked oligosaccharide biosynthetic process
response to salt stress
leaf development
glucosyltransferase activity
dolichyl pyrophosphate Glc2Man9GlcNAc2 alpha-1,2-glucosyltransferase activity

Back To Top

Related Products

  • TaqMan® Fast Advanced Master Mix
  • SuperScript® VILO™ cDNA Synthesis Kit

Your items have has been added!


Host server : magellan-search-5fc94fc45-hqqk4:80/100.66.128.142:80.
git-commit: 7a6627b71d15189168043ee5c63248cc100e5965
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.37.0-2025.10.41-1.0