This product has been added to your favorites list. Go to My Favorites

System Message

OKCancel
LOADING ...
  • Home
  • › Search Tool
  • › Search Results
  • › At02173825_g1
See other IPGAM2 GE Assays ›
Gene Symbol:
iPGAM2
Gene Name:
Phosphoglycerate mutase, 2,3-bisphosphoglycerate-independent
Gene Aliases:
2, 3-biphosphoglycerate-independent phosphoglycerate mutase 2, AT3G08590
Chromosome Location:
Chr.3: 2608308 - 2611555 on Build TAIR10.1
Species:
Arabidopsis
Species Specific ID (Flybase ID):
-
Assay ID At02173825_g1
Size
Availability Made To Order
Catalog # 4351372
Price
Your Price
Online offer:
Check your price ›
  • Genomic Map
  • Assay Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

Assay Details

Gene Symbol: iPGAM2
Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
RefSeq NM_111693.3 NP_187471.1 2 - 3 397 63
NM_180211.2 NP_850542.1 1 - 2 343 63
GenBank mRNA AF424615.1 - 2 - 3 258 63
AK316907.1 - 1 - 2 344 63
AY039969.1 - 2 - 3 259 63
AY113910.1 - 1 - 2 169 63

Back To Top

More Information


Set Membership:

Amplicon length less than or equal to 70 Detect Genomic DNA Arabidopsis thaliana Probe spans exons

Panther Classification:

Molecular Function -

mutase

Gene Ontology Categories:

Function(s) Process(es)

glucose catabolic process
glycolytic process
pollen development
response to blue light
response to abscisic acid
response to carbon dioxide
stomatal movement
cellular carbohydrate metabolic process
response to cadmium ion
manganese ion binding
2,3-bisphosphoglycerate-independent phosphoglycerate mutase activity

Back To Top

Related Products

  • TaqMan® Fast Advanced Master Mix
  • SuperScript® VILO™ cDNA Synthesis Kit

Your items have has been added!


Host server : magellan-search-7db4fb6694-gx648:80/100.66.128.142:80.
git-commit: 672a5a8c50d6b616e949be631a941c86748ade1b
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/search_redesign_header