This product has been added to your favorites list. Go to My Favorites

System Message

OKCancel
LOADING ...
  • Home
  • › Search Tool
  • › Search Results
  • › At02173502_g1
See other RFI2 GE Assays ›
Gene Symbol:
RFI2
Gene Name:
RING/U-box superfamily protein
Gene Aliases:
AT2G47700, F17A22.9, RED AND FAR-RED INSENSITIVE 2
Chromosome Location:
Chr.2: 19552314 - 19554647 on Build TAIR10.1
Species:
Arabidopsis
Species Specific ID (Flybase ID):
-
Assay ID At02173502_g1
Size
Availability Made To Order
Catalog # 4351372
Price 374.00
Your Price
Online offer:
374.0
Check your price ›
  • Genomic Map
  • Assay Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

Assay Details

Gene Symbol: RFI2
Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
RefSeq NM_001337272.1 NP_001324431.1 1 - 2 423 84
NM_180147.3 NP_850478.1 1 - 2 486 84
GenBank mRNA BT002412.1 - 1 - 2 389 84
BT008845.1 - 1 - 2 190 84
DQ086859.1 - 1 - 2 190 84

Back To Top

More Information


Set Membership:

Amplicon length between 71 and 85 Detect Genomic DNA Arabidopsis thaliana Probe spans exons

Gene Ontology Categories:

Function(s) Process(es)

circadian rhythm
response to red light
response to far red light
regulation of gene expression
negative regulation of gene expression
protein ubiquitination
regulation of red or far-red light signaling pathway
regulation of photoperiodism, flowering
ubiquitin-protein transferase activity
protein binding

Back To Top

Related Products

  • TaqMan® Fast Advanced Master Mix
  • SuperScript® VILO™ cDNA Synthesis Kit

Your items have has been added!


Host server : magellan-search-5fc94fc45-4mbwj:80/100.66.130.94:80.
git-commit: 7a6627b71d15189168043ee5c63248cc100e5965
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.37.0-2025.10.41-1.0