This product has been added to your favorites list. Go to My Favorites

System Message

OKCancel
LOADING ...
  • Home
  • › Search Tool
  • › Search Results
  • › At02172922_g1
See other SPPL3 GE Assays ›
Gene Symbol:
SPPL3
Gene Name:
SIGNAL PEPTIDE PEPTIDASE-LIKE 3
Gene Aliases:
ARABIDOPSIS THALIANA SIGNAL PEPTIDE PEPTIDASE-LIKE 3, AT2G43070, ATSPPL3, SIGNAL PEPTIDE PEPTIDASE-LIKE 3
Chromosome Location:
Chr.2: 17910523 - 17915009 on Build TAIR10.1
Species:
Arabidopsis
Species Specific ID (Flybase ID):
-
Assay ID At02172922_g1
Best Coverage
Size
Availability Made To Order
Catalog # 4351372
Price
Your Price
Online offer:
Check your price ›
  • Genomic Map
  • Assay Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

Assay Details

Gene Symbol: SPPL3
Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
RefSeq NM_001337011.1 NP_001318409.1 9 - 10 1233 65
NM_001337012.1 NP_001325361.1 9 - 10 1293 65
NM_001337013.1 NP_001325362.1 9 - 10 1326 65
NM_180052.4 NP_850383.3 9 - 10 1352 65
GenBank mRNA AK229438.1 - 9 - 10 1194 65
BT023438.1 - 9 - 10 1145 65

Back To Top

More Information


Set Membership:

Amplicon length less than or equal to 70 Detect Genomic DNA Recommended Assay Arabidopsis thaliana Probe spans exons

Panther Classification:

Molecular Function -

aspartic protease

Gene Ontology Categories:

Function(s) Process(es)

membrane protein proteolysis
aspartic-type endopeptidase activity
aspartic endopeptidase activity, intramembrane cleaving

Back To Top

Related Products

  • TaqMan® Fast Advanced Master Mix
  • SuperScript® VILO™ cDNA Synthesis Kit

Your items have has been added!


Host server : magellan-search-5fc94fc45-hqqk4:80/100.66.128.142:80.
git-commit: 7a6627b71d15189168043ee5c63248cc100e5965
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.37.0-2025.10.41-1.0