This product has been added to your favorites list. Go to My Favorites

System Message

OKCancel
LOADING ...
  • Home
  • › Search Tool
  • › Search Results
  • › At02171873_g1
See other LYM2 GE Assays ›
Gene Symbol:
LYM2
Gene Name:
lysm domain GPI-anchored protein 2 precursor
Gene Aliases:
AT2G17120, CEBiP-like 1, CL-1, F6P23.25, F6P23_25, LYP1, LysM-containing receptor protein 1, lysM domain GPI-anchored protein 2 precursor, lysm domain GPI-anchored protein 2 precursor
Chromosome Location:
Chr.2: 7458930 - 7460819 on Build TAIR10.1
Species:
Arabidopsis
Species Specific ID (Flybase ID):
-
Assay ID At02171873_g1
Best Coverage
Size
Availability Made To Order
Catalog # 4351372
Price
Your Price
Online offer:
Check your price ›
  • Genomic Map
  • Assay Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

Assay Details

Gene Symbol: LYM2
Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
RefSeq NM_127266.4 NP_565406.1 1 - 2 877 81
GenBank mRNA AF428464.1 - 1 - 2 694 81
AY056221.1 - 1 - 2 666 81
AY125560.1 - 1 - 2 651 81

Back To Top

More Information


Set Membership:

Amplicon length between 71 and 85 Detect Genomic DNA Recommended Assay Arabidopsis thaliana Probe spans exons

Gene Ontology Categories:

Process(es)

chitin binding

Back To Top

Related Products

  • TaqMan® Fast Advanced Master Mix
  • SuperScript® VILO™ cDNA Synthesis Kit

Your items have has been added!


Host server : magellan-search-5fc94fc45-hqqk4:80/100.66.128.142:80.
git-commit: 7a6627b71d15189168043ee5c63248cc100e5965
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.37.0-2025.10.41-1.0