This product has been added to your favorites list. Go to My Favorites

System Message

OKCancel
LOADING ...
  • Home
  • › Search Tool
  • › Search Results
  • › At02168928_g1
See other DIS2 GE Assays ›
Gene Symbol:
DIS2
Gene Name:
Arp2/3 complex, 34 kD subunit p34-Arc
Gene Aliases:
ACTIN-RELATED PROTEIN C2A, ARPC2A, AT1G30825, DISTORTED TRICHOMES 2
Chromosome Location:
Chr.1: 10960908 - 10963231 on Build TAIR10.1
Species:
Arabidopsis
Species Specific ID (Flybase ID):
-
Assay ID At02168928_g1
Size
Availability Made To Order
Catalog # 4351372
Price 374.00
Your Price
Online offer:
374.0
Check your price ›
  • Genomic Map
  • Assay Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

Assay Details

Gene Symbol: DIS2
Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
RefSeq NM_102820.3 NP_564364.1 7 - 8 997 94
GenBank mRNA AK118475.1 - 7 - 8 961 94
AY084256.1 - 7 - 8 961 94
BT005308.1 - 7 - 8 799 94

Back To Top

More Information


Set Membership:

Amplicon length between 86 and 100 Detect Genomic DNA Arabidopsis thaliana Probe spans exons

Gene Ontology Categories:

Function(s) Process(es)

actin filament organization
trichome morphogenesis
actin filament polymerization
Arp2/3 complex-mediated actin nucleation
structural constituent of cytoskeleton
protein binding
actin filament binding

Back To Top

Related Products

  • TaqMan® Fast Advanced Master Mix
  • SuperScript® VILO™ cDNA Synthesis Kit

Your items have has been added!


Host server : magellan-search-56ffd697dc-88d4p:80/100.66.130.94:80.
git-commit: 082a6d415be982b1f92c35e55dd71609d4576d3e
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.36.1-2025.09.37-1.0