This product has been added to your favorites list. Go to My Favorites

System Message

OKCancel
LOADING ...
  • Home
  • › Search Tool
  • › Search Results
  • › At02166783_g1
See other MGT3 GE Assays ›
Gene Symbol:
MGT3
Gene Name:
magnesium transporter 3
Gene Aliases:
AT2G03620, AtMGT3, AtMRS2-5, F19B11.7, F19B11_7, MRS2-5, magnesium transporter 3
Chromosome Location:
Chr.2: 1100325 - 1103495 on Build TAIR10.1
Species:
Arabidopsis
Species Specific ID (Flybase ID):
-
Assay ID At02166783_g1
Size
Availability Made To Order
Catalog # 4351372
Price 374.00
Your Price
Online offer:
374.0
Check your price ›
  • Genomic Map
  • Assay Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

Assay Details

Gene Symbol: MGT3
Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
RefSeq NM_001124787.1 NP_001118259.1 6 - 7 1530 136
NM_126412.6 NP_178460.1 5 - 6 1204 136
GenBank mRNA AF499434.1 - 4 - 5 993 136
AY070380.1 - 5 - 6 1125 136
AY096567.1 - 4 - 5 982 136

Back To Top

More Information


Set Membership:

Amplicon length greater than or equal to 101 Detect Genomic DNA Arabidopsis thaliana Probe spans exons

Panther Classification:

Molecular Function -

RNA splicing factor

Gene Ontology Categories:

Function(s) Process(es)

magnesium ion transport
magnesium ion transmembrane transport
magnesium ion transmembrane transporter activity

Back To Top

Related Products

  • TaqMan® Fast Advanced Master Mix
  • SuperScript® VILO™ cDNA Synthesis Kit

Your items have has been added!


Host server : magellan-search-5fc94fc45-hqqk4:80/100.66.128.142:80.
git-commit: 7a6627b71d15189168043ee5c63248cc100e5965
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.37.0-2025.10.41-1.0