This product has been added to your favorites list. Go to My Favorites

System Message

OKCancel
LOADING ...
  • Home
  • › Search Tool
  • › Search Results
  • › At02165064_g1
See other CRK29 GE Assays ›
Gene Symbol:
CRK29
Gene Name:
cysteine-rich RLK (RECEPTOR-like protein kinase) 29
Gene Aliases:
AT4G21410, F18E5.30, cysteine-rich RLK (RECEPTOR-like protein kinase) 29
Chromosome Location:
Chr.4: 11401711 - 11405235 on Build TAIR10.1
Species:
Arabidopsis
Species Specific ID (Flybase ID):
-
Assay ID At02165064_g1
Best Coverage
Size
Availability Made To Order
Catalog # 4351372
Price 374.00
Your Price
Online offer:
374.0
Check your price ›
  • Genomic Map
  • Assay Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

Assay Details

Gene Symbol: CRK29
Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
RefSeq NM_001341488.1 NP_001320020.1 2 - 3 590 71
NM_001341489.1 NP_001329810.1 2 - 3 1175 71
NM_118261.4 NP_193872.2 2 - 3 1175 71
GenBank mRNA AY074844.1 - 2 - 3 996 71

Back To Top

More Information


Set Membership:

Amplicon length between 71 and 85 Detect Genomic DNA Recommended Assay Arabidopsis thaliana Probe spans exons

Panther Classification:

Molecular Function -

transmembrane signal receptor

Gene Ontology Categories:

Function(s) Process(es)

protein phosphorylation
response to abscisic acid
protein kinase activity
protein serine/threonine kinase activity
ATP binding
kinase activity

Back To Top

Related Products

  • TaqMan® Fast Advanced Master Mix
  • SuperScript® VILO™ cDNA Synthesis Kit

Your items have has been added!


Host server : magellan-search-56ffd697dc-88d4p:80/100.66.130.94:80.
git-commit: 082a6d415be982b1f92c35e55dd71609d4576d3e
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.36.1-2025.09.37-1.0