This product has been added to your favorites list. Go to My Favorites

System Message

OKCancel
LOADING ...
  • Home
  • › Search Tool
  • › Search Results
  • › At02163678_g1
See other LCAT3 GE Assays ›
Gene Symbol:
LCAT3
Gene Name:
lecithin:cholesterol acyltransferase 3
Gene Aliases:
ARABIDOPSIS LECITHIN:CHOLESTEROL ACYLTRANSFERASE 3, AT3G03310, ATLCAT3, T21P5.27, T21P5_27, lecithin:cholesterol acyltransferase 3
Chromosome Location:
Chr.3: 778568 - 781703 on Build TAIR10.1
Species:
Arabidopsis
Species Specific ID (Flybase ID):
-
Assay ID At02163678_g1
Size
Availability Made To Order
Catalog # 4351372
Price 374.00
Your Price
Online offer:
374.0
Check your price ›
  • Genomic Map
  • Assay Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

Assay Details

Gene Symbol: LCAT3
Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
RefSeq NM_111202.4 NP_566201.1 5 - 6 841 101
GenBank mRNA AF421148.1 - 5 - 6 626 101
AY054239.1 - 5 - 6 659 101
AY056414.1 - 5 - 6 716 101
AY113174.1 - 5 - 6 626 101

Back To Top

More Information


Set Membership:

Amplicon length greater than or equal to 101 Detect Genomic DNA Arabidopsis thaliana Probe spans exons

Panther Classification:

Molecular Function -

acyltransferase

Gene Ontology Categories:

Process(es)

O-acyltransferase activity
phosphatidylcholine 1-acylhydrolase activity
phosphatidylserine 1-acylhydrolase activity
1-acyl-2-lysophosphatidylserine acylhydrolase activity

Back To Top

Related Products

  • TaqMan® Fast Advanced Master Mix
  • SuperScript® VILO™ cDNA Synthesis Kit

Your items have has been added!


Host server : magellan-search-5c796d7db4-nk9d7:80/100.66.128.142:80.
git-commit: d3b3aa98da0ae1b146c174fddb0766cfa6979daf
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.36.2-2025.09.39-1.0