Assay Details
Gene Symbol: PLPB | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001335131.1 | NP_001318183.1 | 4 - 5 | 1351 | 110 | |
NM_126326.5 | NP_565288.1 | 4 - 5 | 1391 | 110 | ||
NM_179597.1 | NP_849928.1 | 4 - 5 | 1340 | 110 | ||
GenBank mRNA | AB038798.1 | - | 4 - 5 | 1341 | 110 | |
AY045642.1 | - | 4 - 5 | 1143 | 110 | ||
AY058232.1 | - | 4 - 5 | 935 | 110 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() ![]() |
