This product has been added to your favorites list. Go to My Favorites

System Message

OKCancel
LOADING ...
  • Home
  • › Search Tool
  • › Search Results
  • › At02161944_g1
See other STI GE Assays ›
Gene Symbol:
STI
Gene Name:
AAA-type ATPase family protein
Gene Aliases:
AT2G02480, DNA POLYMERASE III-LIKE PROTEIN, GAMMA SUBUNIT, STICHEL, T8K22.22, T8K22_22
Chromosome Location:
Chr.2: 660655 - 665635 on Build TAIR10.1
Species:
Arabidopsis
Species Specific ID (Flybase ID):
-
Assay ID At02161944_g1
Size
Availability Made To Order
Catalog # 4351372
Price 374.00
Your Price
Online offer:
374.0
Check your price ›
  • Genomic Map
  • Assay Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

Assay Details

Gene Symbol: STI
Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
RefSeq NM_001335120.1 NP_001324706.1 6 - 7 3852 107
NM_126303.3 NP_565285.1 6 - 7 3841 107
GenBank mRNA AF264023.1 - 6 - 7 3718 107

Back To Top

More Information


Set Membership:

Amplicon length greater than or equal to 101 3' Most Detect Genomic DNA Arabidopsis thaliana Probe spans exons

Panther Classification:

Molecular Function -

DNA-directed DNA polymerase

Gene Ontology Categories:

Function(s) Process(es)

DNA-dependent DNA replication
DNA repair
trichome differentiation
trichome branching
DNA clamp loader activity
DNA-directed DNA polymerase activity

Back To Top

Related Products

  • TaqMan® Fast Advanced Master Mix
  • SuperScript® VILO™ cDNA Synthesis Kit

Your items have has been added!


Host server : magellan-search-5fc94fc45-4mbwj:80/100.66.130.94:80.
git-commit: 7a6627b71d15189168043ee5c63248cc100e5965
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.37.0-2025.10.41-1.0