This product has been added to your favorites list. Go to My Favorites

System Message

OKCancel
LOADING ...
  • Home
  • › Search Tool
  • › Search Results
  • › At02161805_g1
See other ETL1 GE Assays ›
Gene Symbol:
ETL1
Gene Name:
SNF2 domain-containing protein / helicase domain-containing protein
Gene Aliases:
AT2G02090, CHA19, CHR19, CHROMATIN REMODELING 19, F5O4.14, F5O4_14
Chromosome Location:
Chr.2: 523223 - 527068 on Build TAIR10.1
Species:
Arabidopsis
Species Specific ID (Flybase ID):
-
Assay ID At02161805_g1
Size
Availability Made To Order
Catalog # 4351372
Price 374.00
Your Price
Online offer:
374.0
Check your price ›
  • Genomic Map
  • Assay Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

Assay Details

Gene Symbol: ETL1
Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
RefSeq NM_126270.2 NP_178318.1 2 - 3 959 121
GenBank mRNA AY059857.1 - 2 - 3 898 121
BT002576.1 - 2 - 3 701 121

Back To Top

More Information


Set Membership:

Amplicon length greater than or equal to 101 Detect Genomic DNA Arabidopsis thaliana Probe spans exons

Panther Classification:

Molecular Function -

DNA helicase

Gene Ontology Categories:

Function(s) Process(es)

cellular response to DNA damage stimulus
ATP-dependent chromatin remodeling
cellular response to UV-C
DNA binding
chromatin binding
protein binding
DNA-dependent ATPase activity

Back To Top

Related Products

  • TaqMan® Fast Advanced Master Mix
  • SuperScript® VILO™ cDNA Synthesis Kit

Your items have has been added!


Host server : magellan-search-5fc94fc45-4mbwj:80/100.66.130.94:80.
git-commit: 7a6627b71d15189168043ee5c63248cc100e5965
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.37.0-2025.10.41-1.0