This product has been added to your favorites list. Go to My Favorites

System Message

OKCancel
LOADING ...
  • Home
  • › Search Tool
  • › Search Results
  • › At02160224_g1
See other DIM1B GE Assays ›
Gene Symbol:
DIM1B
Gene Name:
Ribosomal RNA adenine dimethylase family protein
Gene Aliases:
AT5G66360, K1L20.14, K1L20_14, adenosine dimethyl transferase 1B
Chromosome Location:
Chr.5: 26510146 - 26511844 on Build TAIR10.1
Species:
Arabidopsis
Species Specific ID (Flybase ID):
-
Assay ID At02160224_g1
Best Coverage
Size
Availability Made To Order
Catalog # 4351372
Price
Your Price
Online offer:
Check your price ›
  • Genomic Map
  • Assay Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

Assay Details

Gene Symbol: DIM1B
Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
RefSeq NM_126034.3 NP_201437.2 1 - 2 594 93
NM_203274.2 NP_975003.1 1 - 2 596 93
GenBank mRNA AY120713.1 - 1 - 2 513 93
BT008836.1 - 1 - 2 413 93

Back To Top

More Information


Set Membership:

Amplicon length between 86 and 100 3' Most Detect Genomic DNA Recommended Assay Arabidopsis thaliana Probe spans exons

Panther Classification:

Molecular Function -

RNA methyltransferase

Gene Ontology Categories:

Function(s) Process(es)

rRNA modification
rRNA methylation
adenosine metabolic process
rRNA (adenine-N6,N6-)-dimethyltransferase activity

Back To Top

Related Products

  • TaqMan® Fast Advanced Master Mix
  • SuperScript® VILO™ cDNA Synthesis Kit

Your items have has been added!


Host server : magellan-search-5fc94fc45-hqqk4:80/100.66.128.142:80.
git-commit: 7a6627b71d15189168043ee5c63248cc100e5965
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.37.0-2025.10.41-1.0