This product has been added to your favorites list. Go to My Favorites

System Message

OKCancel
LOADING ...
  • Home
  • › Search Tool
  • › Search Results
  • › At02157532_g1
See other CSTF77 GE Assays ›
Gene Symbol:
CSTF77
Gene Name:
Tetratricopeptide repeat (TPR)-like superfamily protein
Gene Aliases:
ARABIDOPSIS THALIANA CLEAVAGE STIMULATION FACTOR 77, AT1G17760, ATCSTF77, F11A6.10, F11A6_10
Chromosome Location:
Chr.1: 6110107 - 6116617 on Build TAIR10.1
Species:
Arabidopsis
Species Specific ID (Flybase ID):
-
Assay ID At02157532_g1
Size
Availability Made To Order
Catalog # 4351372
Price
Your Price
Online offer:
Check your price ›
  • Genomic Map
  • Assay Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

Assay Details

Gene Symbol: CSTF77
Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
RefSeq NM_101639.3 NP_173218.2 5 - 6 737 80
GenBank mRNA BT002320.1 - 5 - 6 391 80

Back To Top

More Information


Set Membership:

Amplicon length between 71 and 85 Detect Genomic DNA Arabidopsis thaliana Probe spans exons

Panther Classification:

Molecular Function -

mRNA polyadenylation factor

Gene Ontology Categories:

Function(s) Process(es)

gene silencing by RNA
RNA 3'-end processing
antisense RNA metabolic process
negative regulation of transcription, DNA-templated
regulation of gene silencing
mRNA binding
protein binding

Back To Top

Related Products

  • TaqMan® Fast Advanced Master Mix
  • SuperScript® VILO™ cDNA Synthesis Kit

Your items have has been added!


Host server : magellan-search-5fc94fc45-hqqk4:80/100.66.128.142:80.
git-commit: 7a6627b71d15189168043ee5c63248cc100e5965
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.37.0-2025.10.41-1.0