This product has been added to your favorites list. Go to My Favorites

System Message

OKCancel
LOADING ...
  • Home
  • › Search Tool
  • › Search Results
  • › At02156056_g1
See other AVP1 GE Assays ›
Gene Symbol:
AVP1
Gene Name:
Inorganic H pyrophosphatase family protein
Gene Aliases:
ARABIDOPSIS THALIANA V-PPASE 3, AT1G15690, ATAVP3, AVP-3, AtAVP1, AtVHP1;1, F7H2.3, F7H2_3, FUGU 5, FUGU5, VACUOLAR H+ - PYROPHOSPHATASE AVP-3, VHP1
Chromosome Location:
Chr.1: 5398926 - 5402733 on Build TAIR10.1
Species:
Arabidopsis
Species Specific ID (Flybase ID):
-
Assay ID At02156056_g1
Size
Availability Made To Order
Catalog # 4351372
Price
Your Price
Online offer:
Check your price ›
  • Genomic Map
  • Assay Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

Assay Details

Gene Symbol: AVP1
Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
RefSeq NM_101437.5 NP_173021.1 6 - 7 2237 62
GenBank mRNA BT002481.1 - 6 - 7 2173 62
M81892.1 - 6 - 7 2203 62

Back To Top

More Information


Set Membership:

Amplicon length less than or equal to 70 Detect Genomic DNA Arabidopsis thaliana Probe spans exons

Gene Ontology Categories:

Function(s) Process(es)

sucrose metabolic process
response to water deprivation
response to salt stress
auxin polar transport
establishment or maintenance of transmembrane electrochemical gradient
leaf development
cell wall pectin metabolic process
hydrogen ion transmembrane transport
regulation of starch metabolic process
mRNA binding
inorganic diphosphatase activity
hydrogen-translocating pyrophosphatase activity

Back To Top

Related Products

  • TaqMan® Fast Advanced Master Mix
  • SuperScript® VILO™ cDNA Synthesis Kit

Your items have has been added!


Host server : magellan-search-69fc894774-tvlwx:80/100.66.128.142:80.
git-commit: 8e82defa9bf25b3b77800b5e306013d4ba941391
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.36.2-2025.09.39-1.0