Thermo Fisher Scientific
Semiconductor manufacturing stories and solutions about a more connected, more autonomous, and smarter world
  • Categories
    • Advancing Materials
    • Advancing Mining
    • AnalyteGuru
    • Analyzing Metals
    • Ask a Scientist
    • Behind the Bench
    • Biotech at Scale
    • Clinical Conversations
    • Examining Food
    • Identifying Threats
    • Illuminating Semiconductors
    • Life in Atomic Resolution
    • Life in the Lab
    • OEMpowered
    • The Connected Lab
  • About Us
  • Contact
Accelerating ScienceIlluminating Semiconductors / Semiconductor / Transmission Electron Microscopy in Semiconductors: Generating Ground Truth Insights
Transmission Electron Microscopy in Semiconductors: Generating Ground Truth Insights
Written by Paul Kirby, Senior Marketing Manager, Thermo Fisher Scientific | Published: 08.14.2025
What is the current state of TEM in semiconductor analysis?
Transmission electron microscopy (TEM) has become a crucial tool in semiconductor manufacturing. Modern semiconductor fabrication requires atomic-scale imaging and analysis capabilities, particularly for complex three-dimensional (3D) structures, such as gate-all-around (GAA) logic transistors and 3D NAND memory channels.
How does TEM technology work?
TEM operates by directing an electron beam through an ultra-thin sample. The transmitted electrons interact with various filters, detectors, and cameras below the sample. Key operational aspects include:
  • Acceleration voltages—60 kV to 300 kV range for semiconductor applications
  • Resolution capability—down to individual atom visualization
  • Analysis modes—TEM and STEM (scanning TEM)
Diagram comparing optical microscope with light source and lenses to TEM with electron source and lenses
Fig 1: Comparison diagram of optical microscope and TEM configurations.
What are the key differences between TEM and STEM modes?
TEM mode:
  • Uses a focused beam on the sample
  • Captures transmitted electrons in a single snapshot
  • Employs high-speed, high-resolution cameras
  • Offers faster, simpler operation
Schematic of TEM mode operation displaying electron source, lenses, specimen holder, and viewing screen in sequence.
Fig 2: Schematic illustration of TEM mode operation.
STEM mode:
  • Utilizes electromagnetic scan coils
  • Scans a focused beam across the sample
  • Multiple detector configurations available
  • Provides flexible, high-contrast imaging
Schematic of STEM mode operation displaying electron source, lenses, scanning coils, specimen holder, detector, and viewing screen in sequence
Fig 3: Schematic illustration of STEM mode operation.
How has automation enhanced TEM analysis?
Modern semiconductor TEM systems feature significant automation improvements:
  • Simplified parameter selection
  • Automated alignment and calibration
  • Reduced data distortion risk
  • Atomic-scale image calibration
  • Streamlined operation for various sample types
Why is TEM critical for semiconductor manufacturing?
TEM serves as the primary source for reference data in semiconductor design and fabrication due to:
  • Increased 3D device complexity
  • Shrinking critical dimensions
  • EUV lithography requirements
  • Need for atomic interface analysis
  • Strain and magnetic field measurements
What are the key applications and the industry impact?
Leading semiconductor manufacturers now generate tens of thousands of TEM datasets monthly for:
  • Process variation analysis
  • Wafer fab tool calibration
  • Defect mechanism investigation
  • Quality control
Line graph depicting the increasing trend in semiconductor TEM analysis usage
Fig 4: Graph showing growth trend in semiconductor TEM analysis usage.
Summary
  • TEM technology provides atomic-level imaging and analysis capabilities
  • Dual-mode operation (TEM/STEM) offers flexible analysis options
  • Automation advances have simplified operation and improved reliability
  • Critical role in semiconductor manufacturing quality control
  • Essential for next-generation device development
Subscribe to Thermo Fisher Scientific's semiconductor updates and resources
//
Paul Kirby is a Senior Marketing Manager at Thermo Fisher Scientific
TEM lamella preparation
High-Quality TEM Lamella Preparation: Critical Factors and Best Practices
Introduction: The quality of transmission electron microscop... by David Akerson / 09.02.2025
Read More
Removing Large Volumes of Material Fast with FIB-SEMs
Removing Large Volumes of Material Fast with FIB-SEMs
Focused ion beam scanning electron microscopes (FIB-SEMs) ar... by David Akerson / 08.29.2025
Read More
Physical and electrical failure analysis for power devices
Physical and Electrical Failure Analysis of Power Semiconductor Devices
World energy consumption is expected to grow by nearly 50% b... by Roger Alvis / 06.18.2025
Read More
Revolutionizing the way TEM metrology data is collected
As I discussed in my 2024 year-end article, the semiconducto... by Mohan Iyer / 03.04.2025
Read More
Paul Kirby
Leave a Reply Cancel reply
Your email address will not be published. Required fields are marked *
Physical and Electrical Failure Analysis of Power Semiconductor Devices
Removing Large Volumes of Material Fast with FIB-SEMs