">
Balance Enclosures
Explore TopAir balance enclosures
TopAir balance enclosures/weighing stations are an excellent solution for the weighing of potent compounds and chemicals and allow precise weighing data up to 4 decimal places. They provide a safe work environment for lab staff working with acids and harsh chemicals through both particulate and liquid chemical filtration, which is enabled through a variety of available filters.
Balance enclosure features
Advanced Operation System
  • Colour 5" touch screen
  • 10-speed fan
  • Hour counter for filter
  • Filter replacement alarm
Custom dimensions available
TopAir's weighing stations can be customised to the requirements of each client.
Balance enclosure specifications
Weighing station
Made of high-quality non-corrosive polypropylene, the cabinets feature a high level of chemical resistance and are easy to clean. Polypropylene increases the product’s tensile strength and improves its thermal characteristics.
Built-in sealed polypropylene worktop or choice of epoxy, stainless steel, ceramic, or Trespa. Tempered glass sliding front sash and station comes in five standard sizes.
View specification
Sizes
5 standard sizes: (external W x D x H)
  • 600 x 750 x 1223 mm
  • 900 x 750 x 1223 mm
  • 1200 x 750 x 1223 mm
  • 1500 x 750 x 1223 mm
  • 1800 x 750 x 1223 mm
  • Custom
Filter HEPA filter with convenient front access for filter replacement
Optional accessories
  • Stand
  • Optional: No worktop, with independent table, or optical table
  • XL model with 2 internal filters (increases height of cabinets by 100 mm)
  • Variety of filters
Air velocity
  • High efficiency quiet EC fan
  • Wind speed at 0.5±0.1 m/s, 100±20 FPM
Lighting Eco-friendly, cost-effective 800 LUX LED lighting separated from the work area
Power supply 230 V, 50/60 Hz, Single phase
Certification EN-14175/CE/ASHRAE 110-1995 certified
">