">
Optimal Centrifuge Speed and Quality
To fit your application
Explore our cold storage solutions
Expertise you can trust when separation matters
We understand the critical role that separations play in your lab's workflow, and we are here to provide you with the expertise and quality you need. With our wide range of centrifuges, innovative rotors, and supporting labware, we offer outstanding performance spin after spin. That is why labs around the world trust their separation tasks to our centrifuges.
5 Reasons why you should choose Thermo Scientific centrifuges
   Trust Thermo Fisher Scientific to deliver outstanding performance spin after spin
  1. Trusted brand with a long history of over 75 years in centrifugation, along with worldwide manufacturing and distribution sites
  2. Developed innovative, high-quality solutions focused on enhancing the user experience, including our state-of-the-art Thermo Scientific™ Fiberlite™ carbon fiber rotors, Auto-Door™ opening, Auto-Lock™ rotor exchange, and ClickSeal™ biocontainment lids, which improve the safety, efficiency, and reproducibility of essential workflows
  3. Comprehensive portfolio of centrifuges and accessories to suit any application or budget, ranging from basic mini centrifuges to industrial floor-standing centrifuges
  4. World-class technical service team providing vital support for installation, service contracts, and compliance services
  5. Focused on meeting the latest safety and sustainability standards—from the idea phase to the final product
Your lab, your application, we have you covered
Academic/Life Science Research
  • Micro & mini centrifuges
  • Compact & clinical centrifuges
  • General purpose centrifuges
  • Ultraspeed centrifuges
  • Superspeed centrifuges
Clinical
  • Micro & mini centrifuges
  • Compact & clinical centrifuges
  • General purpose centrifuges
  • Large capacity centrifuges –    blood banking
Biopharma
  • Micro & mini centrifuges
  • General purpose centrifuges
  • Ultraspeed centrifuges
  • Superspeed centrifuges
  • Large capacity centrifuges - bioprocessing
Bioprocessing
  • Ultraspeed centrifuges
  • Superspeed centrifuges
  • Large capacity centrifuges – bioprocessing
  • Industrial centrifuges
Meet our family of centrifuges that spin around your needs
Benchtop centrifuges – Small in size, big in power
Mini centrifuges
  • Ease of use - Ergonomic design, tool-free rotors, and quickspin options make it fast and effortless
  • Efficient - Ideal for most protocols requiring fast spins  (12,500 rpm / 9,800 x g)
  • Safe - Ensures it will not operate when the lid is open or if a rotor is jammed and operates quietly with low vibration
Micro centrifuges
  • Ease of use - Simplified operation with intuitive controls,  easy-to-read displays and fast one-click centrifuge lid closure
  • Efficient - Fast acceleration and deceleration, and ideal for processing micro-volumes
  • Safe - Ergonomic one-click biocontainment lid opening and closing
Compact centrifuges
  • Ease of use—Secure, push-button Auto-Lock rotor exchange delivers trouble-free  rotor installation 
  • Efficient—Maximised swing-out capacity up to 8 x 50 mL conical tubes. Fixed angle flexibility to spin 50mL conical tubes at over 12,000 x g or microtubes at speeds up to 30,279 x g
  • Safe—Biocontainment sealing options, ClickSeal lids offer glove-friendly, one-handed operation
General purpose centrifuges
  • Ease of use - Improved ergonomics with ClickSeal biocontainment options to improve sample and user safety. Bright, easy-to-use LCD control panel with touchscreen interface
  • Efficient—Auto-Lock rotor exchange provides flexibility to switch between applications in less than 3 seconds without any tools. Ability to hold up to 196 blood tubes, 40 x 50 mL conical tubes, or 24 microplates in one run
  • Safe—Ergonomically designed for improved reach-in during rotor installation
Floor model centrifuges – Maximum flexibility and high throughput
Ultraspeed
centrifuges
  • Efficient - With performance over 1,000,000 x g and up to 150,000 rpm in a compact footprint
  • Versatile - Extensive rotor selection, including unique carbon  fiber rotors, provide application flexibility
  • Simplified - A user-friendly, full-color graphical display and visual sample balancing up to 5 mm keeps operation and programming simple
  • Quiet - Noise level as low as 45 dBA while running at over 100,000 rpm
Superspeed
centrifuges
  • Ergonomics—Carbon fiber rotors provide lightweight alternatives; speed handles for easier and safer lifting of rotors; foot space to allow closer positioning of the user to reduce strain when loading and unloading
  • Safety- Auto-Lock rotor exchange with Auto-ID instant  rotor identification eliminates rotor accidents from improper rotor attachment and overspeeding
  • Sustainability - Energy efficiency settings with Smart Vacuum and Lighthouse mode selection
Blood banking centrifuges
  • Improved capacity - Unique 16 x 500 mL blood bag capacity maximises productivity
  • Ergonomics – Push-button convenience with Auto-Door automatic centrifuge door  opening and closing
  • Simplified - Space-saving design and Auto-Lid instant rotor opening and closing  with lid storage  
  • Quick run set-up and standardisation - Glove-friendly Thermo Scientific™ Centri-Touch™ interface with Thermo Scientific™ Centri-Cross™ function to convert your  existing protocols
Bioprocessing centrifuges
  • Outstanding capacity - Unique 8 x 2000 mL (16 L) capacity  maximises productivity
  • Unique 10 x 1000mL (10L) configuration up to 12,000 x g  with Fiberlite™ carbon fiber F6-10x1000 LEX rotor
  • Ergonomics – Push-button convenience with Auto-Door automatic centrifuge door  opening and closing
  • Simplified - Space-saving design, Auto-Lid instant rotor opening and closing with lid storage, glove-friendly Centri-Touch interface for quick run set up
Centrifuge ware
Select the right vessel and spin with confidence at any scale
There are several factors to consider when selecting the correct vessel for each application: 
  • Chemical compatibility  
  • Volume 
  • Temperature 
  • Relative centrifugal force (RCF) required 
  • Protocols to be used for loading and sample recovery  
  • Cleaning and autoclaving steps
Understanding your requirements before selecting a tube or bottle ensures you make the right choice. Whether your application includes the need for separations, large volume pelleting, protein purification or DNA isolation, the comprehensive selection of Thermo Scientific™ Nalgene™ and Nunc™ centrifuge ware offers a solution for virtually all scales.
Carbon fiber rotors
Thermo Scientific™ Fiberlite™ carbon fiber rotors
Thermo Scientific centrifuges offer an unparalleled selection of rotors. Built to last, the innovative Fiberlite material are lightweight for energy efficiency and resists corrosion and fatigue, reducing the chance of deformity and the need for derating. Fiberlite rotors are backed by a 15-year warranty.  
Explore our full spectrum of centrifuge solutions in 3D
The right equipment spin after spin
We support you with not only world-class products, but also “value beyond product” with our full service offerings and technical support. Click through our curated areas to see how we can support you.
  • Academic
  • Cell culture
  • Biobanking
  • Cell & Gene Therapy
  • Blood processing
  • Bioprocessing
  • Clinical & Diagnostics
Frequently asked questions
Want more information on specific products or workflows?
Feel free to contact us if you have any questions, want more information on specific products and workflows, or wish to discuss your needs. We are always there to partner with you and your lab.
">