Disclaimer

Clicking the images or links will redirect you to a website hosted by BenchSci that provides third-party scientific content. Neither the content nor the BenchSci technology and processes for selection have been evaluated by us; we are providing them as-is and without warranty of any kind, including for use or application of the Thermo Fisher Scientific products presented.

  • Primary Antibodies ›
  • delta Catenin Antibodies

Proteintech

p120 Catenin Monoclonal Antibody (2F7H8), CoraLite® 594

View all (78) delta Catenin antibodies

{{$productOrderCtrl.translations['antibody.pdp.commerceCard.promotion.promotions']}}

{{$productOrderCtrl.translations['antibody.pdp.commerceCard.promotion.viewpromo']}}

{{$productOrderCtrl.translations['antibody.pdp.commerceCard.promotion.promocode']}}: {{promo.promoCode}} {{promo.promoTitle}} {{promo.promoDescription}}. {{$productOrderCtrl.translations['antibody.pdp.commerceCard.promotion.learnmore']}}

Datasheet
Protocols
Questions & Answers
Datasheet
Protocols
Questions & Answers

Cite p120 Catenin Monoclonal Antibody (2F7H8), CoraLite® 594

  • Antibody Testing Data (3)
p120 Catenin Antibody in Immunocytochemistry (ICC/IF)
Group 53 Created with Sketch.
p120 Catenin Antibody in Immunocytochemistry (ICC/IF)
Group 53 Created with Sketch.

FIGURE: 1 / 3

{{ $ctrl.currentElement.advancedVerification.fullName }}
{{ $ctrl.currentElement.advancedVerification.text }}

p120 Catenin Antibody (CL594-66208) in ICC/IF

Immunofluorescent analysis of (-20°C Ethanol) fixed MCF-7 cells using CL594-66208 (p120 Catenin antibody) at dilution of 1:100 {{ $ctrl.currentElement.advancedVerification.fullName }} validation info. View more
Published figure supplied by benchsci-logo
PMID: {{$ctrl.currentElement.benchSciPubmedId}}
View Product

{{$ctrl.videoDesc}}

{{$ctrl.videoLongDesc}}

p120 Catenin Antibody in Immunocytochemistry (ICC/IF)
p120 Catenin Antibody in Immunohistochemistry (PFA fixed) (IHC (PFA))
p120 Catenin Antibody in Immunohistochemistry (PFA fixed) (IHC (PFA))

Product Details

CL594-66208

Applications
Tested Dilution
Publications

Immunohistochemistry (PFA fixed) (IHC (PFA))

1:100
-

Immunocytochemistry (ICC/IF)

1:50-1:500
-
Product Specifications

Species Reactivity

Human, Mouse, Rat

Host/Isotype

Mouse / IgG2b

Class

Monoclonal

Type

Antibody

Clone

2F7H8

Immunogen

Recombinant Protein

Conjugate

CoraLite® 594 CoraLite® 594 CoraLite® 594
  • Unconjugated
  • Request custom conjugation

Excitation/Emission Max

593/614 nm

Form

Liquid

Concentration

2 mg/mL

Purification

Protein A

Storage buffer

PBS, pH 7.3, with 0.5% BSA, 50% glycerol

Contains

0.05% ProClin 300

Storage conditions

-20°C, store in dark

Shipping conditions

Wet ice

Target Information

Delta 1 Catenin is an efficient tyrosine kinase substrate implicated both in cell transformation by SRC and in ligand-induced receptor signaling through the EGF, PDGF, CSF-1 and ERBB2 receptors. The association of catenins to cadherins produces a complex which is linked to the actin filament network, and which seems to be of primary importance for cadherins cell-adhesion properties.

For Research Use Only. Not for use in diagnostic procedures. Not for resale without express authorization.

References (0)

Have you cited this product in a publication?
Let us know so we can reference it here.
Cite this product

Bioinformatics

Protein Aliases: Cadherin-associated Src substrate; Cadherin-associated Src substrate (CAS); CAS; Catenin (cadherin associated protein) delta 1; catenin (cadherin-associated protein), delta 1; Catenin delta-1; catenin src; p120 catenin; P120 CTN; P120 p120; P120 P120CTN; p120(cas); p120(ctn); p120-catenin

View more View less

Gene Aliases: AA409437; AU019353; CAS; Catns; CTNND; CTNND1; KIAA0384; mKIAA0384; P120; p120(CAS); p120(CTN); P120CAS; P120CTN

View more View less

UniProt ID: (Human) O60716, (Mouse) P30999

View more View less

Entrez Gene ID: (Human) 1500, (Mouse) 12388, (Rat) 311163

View more View less

Function(s)
receptor binding protein binding protein kinase binding cadherin binding cadherin binding involved in cell-cell adhesion protein phosphatase binding protein domain specific binding cell adhesion molecule binding
Process(es)
morphogenesis of a polarized epithelium transcription, DNA-templated regulation of transcription, DNA-templated cell adhesion salivary gland morphogenesis Wnt signaling pathway single organismal cell-cell adhesion keratinocyte differentiation epithelial cell differentiation involved in salivary gland development negative regulation of canonical Wnt signaling pathway brain development adherens junction organization
It has to be done as per old AB suggested Products section.
guarantee_icon

Performance Guarantee

If an Invitrogen™ antibody doesn't perform as described on our website or datasheet,we'll replace the product at no cost to you, or provide you with a credit for a future purchase.*

Learn more
help_icon

We're here to help

Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.

Contact tech support

Your items have has been added!


Host server : magellan-search-9dd846b7f-s9zn4:80/100.66.128.142:80.
git-commit: 17e8f6c92ed407c129a469238c057b56fdf8abbb
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.36.2-2025.09.39-1.0