Abnova
{{$productOrderCtrl.translations['antibody.pdp.commerceCard.promotion.promotions']}}
{{$productOrderCtrl.translations['antibody.pdp.commerceCard.promotion.viewpromo']}}
{{$productOrderCtrl.translations['antibody.pdp.commerceCard.promotion.promocode']}}: {{promo.promoCode}} {{promo.promoTitle}} {{promo.promoDescription}}. {{$productOrderCtrl.translations['antibody.pdp.commerceCard.promotion.learnmore']}}
Product may be used with Western Blot (Transfected lysate).
The Isotype of this product is composed of an IgG Mixture.
Immunogen sequence: MPSLRTRREE AEMELSVPGP SPWTPAAQAR VRDAPAVTHP GSAACGTPCC SDTELEAICP HYQQPDCDTR TEDKEFLHKE DIHEDLESQA EISENYAGDV SQVPELGDLC DDVSERDWGV PEGRRLPQSL SQEGDFTPAA MGLLRGPLGE KDLDCNGFDS RFSLSPNLMA CQEIPTEERP HPYDMGGQSF QHSVDLTGHE GVPTAESPLI CNECGKTFQG NPDLIQRQIV HTGEASFMCD DCGKTFSQNS VLKNRHRSHM SEKAYQCSEC GKAFRGHSDF SRHQSHHSSE RPYMCNECGK AFSQNSSLKK HQKSHMSEKP YECNECGKAF RRSSNLIQHQ RIHSGEKPYV CSECGKAFRR SSNLIKHHRT HTGEKPFECG ECGKAFSQSA HLRKHQRVHT GEKPYECNDC GKPFSRVSNL IKHHRVHTGE KPYKCSDCGK AFSQSSSLIQ HRRIHTGEKP HVCNVCGKAF SYSSVLRKHQ IIHTGEKPYR CSVCGKAFSH SSALIQHQGV HTGDKPYACH ECGKTFGRSS NLILHQRVHT GEKPYECTEC GKTFSQSSTL IQHQRIHNGL KPHECNQCGK AFNRSSNLIH HQKVHTGEKP YTCVECGKGF SQSSHLIQHQ IIHTGERPYK CSECGKAFSQ RSVLIQHQRI HTGVKPYDCA ACGKAFSQRS KLIKHQLIHT RE
ZNF16 belongs to the krueppel C2H2-type zinc-finger protein family. It contains 17 C2H2-type zinc fingers. ZNF16 may be involved in transcriptional regulation.The protein encoded by this gene contains a C2H2 type of zinc finger, and thus may function as a transcription factor. This gene is located in a region close to ZNF7/KOX4, a gene also encoding a zinc finger protein, on chromosome 8. Two alternatively spliced variants, encoding the same protein, have been identified.
For Research Use Only. Not for use in diagnostic procedures. Not for resale without express authorization.
Protein Aliases: Zinc finger protein 16; Zinc finger protein KOX9; zinc finger protein Kruppel type 9
Gene Aliases: HZF1; KOX9; ZNF16
UniProt ID: (Human) P17020
Entrez Gene ID: (Human) 7564
If an Invitrogen™ antibody doesn't perform as described on our website or datasheet,we'll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Learn moreGet expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.
Contact tech support