Disclaimer

Clicking the images or links will redirect you to a website hosted by BenchSci that provides third-party scientific content. Neither the content nor the BenchSci technology and processes for selection have been evaluated by us; we are providing them as-is and without warranty of any kind, including for use or application of the Thermo Fisher Scientific products presented.

  • Primary Antibodies ›
  • ZMYM3 Antibodies

NeoBiotechnologies

ZMYM3 Monoclonal Antibody (PCRP-ZMYM3-2F10)

View all (8) ZMYM3 antibodies

{{$productOrderCtrl.translations['antibody.pdp.commerceCard.promotion.promotions']}}

{{$productOrderCtrl.translations['antibody.pdp.commerceCard.promotion.viewpromo']}}

{{$productOrderCtrl.translations['antibody.pdp.commerceCard.promotion.promocode']}}: {{promo.promoCode}} {{promo.promoTitle}} {{promo.promoDescription}}. {{$productOrderCtrl.translations['antibody.pdp.commerceCard.promotion.learnmore']}}

Datasheet
Protocols
Questions & Answers
Datasheet
Protocols
Questions & Answers

Cite ZMYM3 Monoclonal Antibody (PCRP-ZMYM3-2F10)

  • Antibody Testing Data (5)
ZMYM3 Antibody in Immunocytochemistry (ICC/IF)
Group 53 Created with Sketch.
ZMYM3 Antibody in Immunocytochemistry (ICC/IF)
Group 53 Created with Sketch.

FIGURE: 1 / 5

{{ $ctrl.currentElement.advancedVerification.fullName }}
{{ $ctrl.currentElement.advancedVerification.text }}

ZMYM3 Antibody (9203-MSM2-P1) in ICC/IF

Immunocytochemistry/Immunofluorescence analysis of PFA-fixed HeLa cells using ZMYM3 Monoclonal Antibody (Product # 9203-MSM2-P1) followed by goat anti-mouse IgG-CF488 (green). CF640A phalloidin (red).. {{ $ctrl.currentElement.advancedVerification.fullName }} validation info. View more
Published figure supplied by benchsci-logo
PMID: {{$ctrl.currentElement.benchSciPubmedId}}
View Product

{{$ctrl.videoDesc}}

{{$ctrl.videoLongDesc}}

ZMYM3 Antibody in Immunocytochemistry (ICC/IF)
ZMYM3 Antibody in Immunohistochemistry (Paraffin) (IHC (P))
ZMYM3 Antibody in Flow Cytometry (Flow)
ZMYM3 Antibody in Flow Cytometry (Flow)
ZMYM3 Antibody in Peptide array (ARRAY)
ZMYM3 Monoclonal Antibody (PCRP-ZMYM3-2F10)

Product Details

9203-MSM2-P1

Applications
Tested Dilution
Publications

Western Blot (WB)

1-2 µg/mL
-

Immunohistochemistry (Paraffin) (IHC (P))

1-2 µg/mL
-

Immunocytochemistry (ICC/IF)

1-2 µg/mL
-

Flow Cytometry (Flow)

1-2 µg/10^6 cells
-

Peptide Array (Array)

Assay-dependent
-
Product Specifications

Species Reactivity

Human

Host/Isotype

Mouse / IgG2a

Class

Monoclonal

Type

Antibody

Clone

PCRP-ZMYM3-2F10

Immunogen

Recombinant full-length human ZMYM3 protein
View immunogen

Conjugate

Unconjugated Unconjugated Unconjugated

Form

Liquid

Concentration

200 µg/mL

Purification

Protein A/G

Storage buffer

PBS, pH 7.4, with 0.05% BSA

Contains

0.05% sodium azide

Storage conditions

4°C

Shipping conditions

Ambient (domestic); Wet ice (international)

Product Specific Information

Immunohistochemistry (PFA fixed): incubate antibody for 30 min RT. Staining of formalin-fixed tissues requires heating tissue sections in 10mM Tris with 1mM EDTA, pH 9.0, for 45 min at 95 degrees C followed by cooling at RT for 20 minutes.

Target Information

This gene is located on the X chromosome and is subject to X inactivation. It is highly conserved in vertebrates and most abundantly expressed in the brain. The encoded protein is a component of histone deacetylase-containing multiprotein complexes that function through modifying chromatin structure to keep genes silent. A chromosomal translocation (X;13) involving this gene is associated with X-linked cognitive disability. Several alternatively spliced transcript variants have been found for this gene.

For Research Use Only. Not for use in diagnostic procedures. Not for resale without express authorization.

References (0)

Have you cited this product in a publication?
Let us know so we can reference it here.
Cite this product

Bioinformatics

Protein Aliases: Zinc finger MYM-type protein 3; Zinc finger protein 261; zinc finger, MYM-type 3

View more View less

Gene Aliases: DXS6673E; KIAA0385; MYM; XFIM; ZMYM3; ZNF198L2; ZNF261

View more View less

UniProt ID: (Human) Q14202

View more View less

Entrez Gene ID: (Human) 9203

View more View less

Function(s)
RNA polymerase II transcription factor activity, sequence-specific DNA binding DNA binding zinc ion binding
Process(es)
regulation of transcription from RNA polymerase II promoter cytoskeleton organization multicellular organism development regulation of cell morphogenesis
It has to be done as per old AB suggested Products section.
guarantee_icon

Performance Guarantee

If an Invitrogen™ antibody doesn't perform as described on our website or datasheet,we'll replace the product at no cost to you, or provide you with a credit for a future purchase.*

Learn more
help_icon

We're here to help

Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.

Contact tech support

Your items have has been added!


Host server : magellan-search-585975bbdc-x5cgg:80/100.66.128.25:80.
git-commit: a17be567db00e29ba041ba55be3678c43488a11a
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.37.0-2025.10.41-1.0