Disclaimer

Clicking the images or links will redirect you to a website hosted by BenchSci that provides third-party scientific content. Neither the content nor the BenchSci technology and processes for selection have been evaluated by us; we are providing them as-is and without warranty of any kind, including for use or application of the Thermo Fisher Scientific products presented.

  • Primary Antibodies ›
  • AMER1 Antibodies

Proteintech

WTX Polyclonal Antibody

View all (6) AMER1 antibodies

{{$productOrderCtrl.translations['antibody.pdp.commerceCard.promotion.promotions']}}

{{$productOrderCtrl.translations['antibody.pdp.commerceCard.promotion.viewpromo']}}

{{$productOrderCtrl.translations['antibody.pdp.commerceCard.promotion.promocode']}}: {{promo.promoCode}} {{promo.promoTitle}} {{promo.promoDescription}}. {{$productOrderCtrl.translations['antibody.pdp.commerceCard.promotion.learnmore']}}

Datasheet
Protocols
Questions & Answers
Datasheet
Protocols
Questions & Answers

Cite WTX Polyclonal Antibody

  • Antibody Testing Data (4)
WTX Antibody in Immunohistochemistry (Paraffin) (IHC (P))
Group 53 Created with Sketch.
WTX Antibody in Immunohistochemistry (Paraffin) (IHC (P))
Group 53 Created with Sketch.

FIGURE: 1 / 4

{{ $ctrl.currentElement.advancedVerification.fullName }}
{{ $ctrl.currentElement.advancedVerification.text }}

WTX Antibody (55432-1-AP) in IHC (P)

Immunohistochemistry of paraffin-embedded mouse stomach tissue slide using 55432-1-AP (WTX antibody at dilution of 1:200 (under 40x lens). {{ $ctrl.currentElement.advancedVerification.fullName }} validation info. View more
Published figure supplied by benchsci-logo
PMID: {{$ctrl.currentElement.benchSciPubmedId}}
View Product

{{$ctrl.videoDesc}}

{{$ctrl.videoLongDesc}}

WTX Antibody in Immunohistochemistry (Paraffin) (IHC (P))
WTX Antibody in Immunohistochemistry (Paraffin) (IHC (P))
WTX Antibody in Immunohistochemistry (Paraffin) (IHC (P))
WTX Antibody in Immunohistochemistry (Paraffin) (IHC (P))

Product Details

55432-1-AP

Applications
Tested Dilution
Publications

Immunohistochemistry (Paraffin) (IHC (P))

1:20-1:200
-
Product Specifications

Species Reactivity

Human, Mouse

Host/Isotype

Rabbit / IgG

Class

Polyclonal

Type

Antibody

Immunogen

Peptide

Conjugate

Unconjugated Unconjugated Unconjugated

Form

Liquid

Concentration

0.27 mg/mL

Purification

Antigen affinity chromatography

Storage buffer

PBS, pH 7.3, with 50% glycerol

Contains

0.02% sodium azide

Storage conditions

-20°C

Shipping conditions

Wet ice (domestic); Dry ice (international)

Target Information

WTX is involved in kidney development.

For Research Use Only. Not for use in diagnostic procedures. Not for resale without express authorization.

References (0)

Have you cited this product in a publication?
Let us know so we can reference it here.
Cite this product

Bioinformatics

Protein Aliases: adenomatous polyposis coli membrane recruitment 1; Amer1; APC membrane recruitment protein 1; family with sequence similarity 123, member B; family with sequence similarity 123B; Protein FAM123B; RP11-403E24.2; Wilms tumor gene on the X chromosome protein; Wilms tumor on the X

View more View less

Gene Aliases: 2810002O09Rik; AMER1; AW492303; FAM123B; OSCS; WTX

View more View less

UniProt ID: (Human) Q5JTC6, (Mouse) Q7TS75

View more View less

Entrez Gene ID: (Human) 139285, (Mouse) 72345

View more View less

Function(s)
protein binding phosphatidylinositol-4,5-bisphosphate binding beta-catenin binding beta-catenin destruction complex binding lipid binding
Process(es)
Wnt signaling pathway positive regulation of protein ubiquitination proteasome-mediated ubiquitin-dependent protein catabolic process anatomical structure development bone development adipose tissue development regulation of canonical Wnt signaling pathway mesenchymal cell differentiation involved in kidney development negative regulation of canonical Wnt signaling pathway positive regulation of canonical Wnt signaling pathway positive regulation of cellular protein catabolic process beta-catenin destruction complex assembly beta-catenin destruction complex disassembly kidney development
It has to be done as per old AB suggested Products section.
guarantee_icon

Performance Guarantee

If an Invitrogen™ antibody doesn't perform as described on our website or datasheet,we'll replace the product at no cost to you, or provide you with a credit for a future purchase.*

Learn more
help_icon

We're here to help

Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.

Contact tech support

Your items have has been added!


Host server : magellan-search-649bfd9c6b-jlsz7:80/100.66.128.142:80.
git-commit: 6001d82975f0049f9efc09ec70b1b2d89229eeb3
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.36.2-2025.09.39-1.0