Abnova
{{$productOrderCtrl.translations['antibody.pdp.commerceCard.promotion.promotions']}}
{{$productOrderCtrl.translations['antibody.pdp.commerceCard.promotion.viewpromo']}}
{{$productOrderCtrl.translations['antibody.pdp.commerceCard.promotion.promocode']}}: {{promo.promoCode}} {{promo.promoTitle}} {{promo.promoDescription}}. {{$productOrderCtrl.translations['antibody.pdp.commerceCard.promotion.learnmore']}}
Product may be used with Western Blot (Transfected lysate).
The Isotype of this product is composed of an IgG Mixture.
Immunogen sequence: MQRSRAGADE AALLLAGLAL RELEPGCGSP GRGRRGPRPG PGDEAAPALG RRGKGSGGPE AGADGLSRGE RGPRRAAVPE LSAQPAGSPR ASLAGSDGGG GGGSARSSGI SLGYDQRHGS PRSGRSDPRP GPGPPSVGSA RSSVSSLGSR GSAGAYADFL PPGACPAPAR SPEPAGPAPF PLPALPLPPG REGGPSAAER RLEALTRELE RALEARTARD YFGICIKCGL GIYGAQQACQ AMGSLYHTDC FTCDSCGRRL RGKAFYNVGE KVYCQEDFLY SGFQQTADKC SVCGHLIMEM ILQALGKSYH PGCFRCSVCN ECLDGVPFTV DVENNIYCVR DYHTVFAPKC ASCARPILPA QGCETTIRVV SMDRDYHVAC YHCEDCGLQL SGEEGRRCYP LAGHLLCRRC HLRRLQPGPL PSPTVHVTEL
Adapter or scaffold protein which participates in the assembly of numerous protein complexes and is involved in several cellular processes such as cell fate determination, cytoskeletal organization, repression of gene transcription, cell-cell adhesion, cell differentiation, proliferation and migration. Positively regulates microRNA (miRNA)-mediated gene silencing. Negatively regulates Hippo signaling pathway and antagonizes phosphorylation of YAP1. Acts as a transcriptional corepressor for SNAI1 and SNAI2/SLUG-dependent repression of E-cadherin transcription. Acts as a hypoxic regulator by bridging an association between the prolyl hydroxylases and VHL enabling efficient degradation of HIF1A. In podocytes, may play a role in the regulation of actin dynamics and/or foot process cytoarchitecture. In the course of podocyte injury, shuttles into the nucleus and acts as a transcription regulator that represses WT1-dependent transcription regulation, thereby translating changes in slit diaphragm structure into altered gene expression and a less differentiated phenotype. Involved in the organization of the basal body. Involved in cilia growth and positioning. [UniProt]
For Research Use Only. Not for use in diagnostic procedures. Not for resale without express authorization.
Protein Aliases: Wilms tumor protein 1-interacting protein; WT1-interacting protein
Gene Aliases: RGD1306411; WTIP
UniProt ID: (Human) A6NIX2
Entrez Gene ID: (Human) 126374, (Rat) 361552
If an Invitrogen™ antibody doesn't perform as described on our website or datasheet,we'll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Learn moreGet expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.
Contact tech support