Disclaimer

Clicking the images or links will redirect you to a website hosted by BenchSci that provides third-party scientific content. Neither the content nor the BenchSci technology and processes for selection have been evaluated by us; we are providing them as-is and without warranty of any kind, including for use or application of the Thermo Fisher Scientific products presented.

  • Primary Antibodies ›
  • VHL Antibodies

OriGene

VHL Monoclonal Antibody (OTI1E1), TrueMAB™

View all (22) VHL antibodies

{{$productOrderCtrl.translations['antibody.pdp.commerceCard.promotion.promotions']}}

{{$productOrderCtrl.translations['antibody.pdp.commerceCard.promotion.viewpromo']}}

{{$productOrderCtrl.translations['antibody.pdp.commerceCard.promotion.promocode']}}: {{promo.promoCode}} {{promo.promoTitle}} {{promo.promoDescription}}. {{$productOrderCtrl.translations['antibody.pdp.commerceCard.promotion.learnmore']}}

Datasheet
Protocols
Questions & Answers
Datasheet
Protocols
Questions & Answers

Cite VHL Monoclonal Antibody (OTI1E1), TrueMAB™

  • Antibody Testing Data (8)
VHL Antibody in Immunohistochemistry (Paraffin) (IHC (P))
Group 53 Created with Sketch.
VHL Antibody in Immunohistochemistry (Paraffin) (IHC (P))
Group 53 Created with Sketch.

FIGURE: 1 / 8

{{ $ctrl.currentElement.advancedVerification.fullName }}
{{ $ctrl.currentElement.advancedVerification.text }}

VHL Antibody (CF506222) in IHC (P)

Immunohistochemical staining of paraffin-embedded human endometrium tissue within the normal limits using anti-VHL mouse monoclonal antibody. (Heat-induced epitope retrieval by 10mM citric buffer, pH6.0, 120°C for 3min, TA506222) {{ $ctrl.currentElement.advancedVerification.fullName }} validation info. View more
Published figure supplied by benchsci-logo
PMID: {{$ctrl.currentElement.benchSciPubmedId}}
View Product

{{$ctrl.videoDesc}}

{{$ctrl.videoLongDesc}}

VHL Antibody in Immunohistochemistry (Paraffin) (IHC (P))
VHL Antibody in Immunohistochemistry (Paraffin) (IHC (P))
VHL Antibody in Immunohistochemistry (Paraffin) (IHC (P))
VHL Antibody in Immunohistochemistry (Paraffin) (IHC (P))
VHL Antibody in Immunohistochemistry (Paraffin) (IHC (P))
VHL Antibody in Immunohistochemistry (Paraffin) (IHC (P))
VHL Antibody in Immunohistochemistry (Paraffin) (IHC (P))
VHL Antibody in Western Blot (WB)
VHL Monoclonal Antibody (OTI1E1), TrueMAB™

Product Details

CF506222

Applications
Tested Dilution
Publications

Western Blot (WB)

1:4,000
-

Immunohistochemistry (Paraffin) (IHC (P))

1:150
-
Product Specifications

Species Reactivity

Human

Host/Isotype

Mouse / IgG2b

Class

Monoclonal

Type

Antibody

Clone

OTI1E1

Immunogen

Full length human recombinant protein of human VHL produced in HEK293T cell.
View immunogen

Conjugate

Unconjugated Unconjugated Unconjugated

Form

Lyophilized

Concentration

1 mg/mL

Purification

Affinity chromatography

Storage buffer

PBS, pH 7.3, with 8% trehalose

Contains

no preservative

Storage conditions

-20°C, Avoid Freeze/Thaw Cycles

Shipping conditions

Ambient (domestic); Wet ice (international)

Product Specific Information

For reconstitution, we recommend adding 100 µL distilled water to a final antibody concentration of about 1 mg/mL. To use this carrier-free antibody for conjugation experiments, we strongly recommend performing another round of desalting. (Zeba Spin Desalting Columns, 7KMWCO, 0.5 mL, Product # 89882)

Target Information

Von Hippel-Lindau syndrome (VHL) is a dominantly inherited familial cancer syndrome predisposing to a variety of malignant and benign tumors. A germline mutation of this gene is the basis of familial inheritance of VHL syndrome. The protein encoded by this gene is a component of the protein complex that includes elongin B, elongin C, and cullin-2, and possesses ubiquitin ligase E3 activity. This protein is involved in the ubiquitination and degradation of hypoxia-inducible-factor (HIF), which is a transcription factor that plays a central role in the regulation of gene expression by oxygen. RNA polymerase II subunit POLR2G/RPB7 is also reported to be a target of this protein. Alternatively spliced transcript variants encoding distinct isoforms have been observed.

For Research Use Only. Not for use in diagnostic procedures. Not for resale without express authorization.

References (0)

Have you cited this product in a publication?
Let us know so we can reference it here.
Cite this product

Bioinformatics

Protein Aliases: elongin binding protein; Protein G7; pVHL; VHL19; VHLp18(MEA); Von Hippel Lindau; von hippel-lindau; von Hippel-Lindau disease tumor suppressor; von Hippel-Lindau tumor suppressor, E3 ubiquitin protein ligase

View more View less

Gene Aliases: HRCA1; pVHL; RCA1; VHL; VHL1

View more View less

UniProt ID: (Human) P40337

View more View less

Entrez Gene ID: (Human) 7428

View more View less

Function(s)
ubiquitin-protein transferase activity protein binding transcription factor binding enzyme binding ubiquitin protein ligase activity
Process(es)
negative regulation of transcription from RNA polymerase II promoter cell morphogenesis regulation of transcription, DNA-templated proteolysis negative regulation of cell proliferation protein ubiquitination negative regulation of apoptotic process positive regulation of cell differentiation positive regulation of transcription, DNA-templated protein stabilization regulation of transcription from RNA polymerase II promoter in response to hypoxia negative regulation of transcription from RNA polymerase II promoter in response to hypoxia
It has to be done as per old AB suggested Products section.
guarantee_icon

Performance Guarantee

If an Invitrogen™ antibody doesn't perform as described on our website or datasheet,we'll replace the product at no cost to you, or provide you with a credit for a future purchase.*

Learn more
help_icon

We're here to help

Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.

Contact tech support

Your items have has been added!


Host server : magellan-search-649bfd9c6b-jlsz7:80/100.66.128.142:80.
git-commit: 6001d82975f0049f9efc09ec70b1b2d89229eeb3
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.36.2-2025.09.39-1.0