Disclaimer

Clicking the images or links will redirect you to a website hosted by BenchSci that provides third-party scientific content. Neither the content nor the BenchSci technology and processes for selection have been evaluated by us; we are providing them as-is and without warranty of any kind, including for use or application of the Thermo Fisher Scientific products presented.

  • Primary Antibodies ›
  • VEGF Receptor 3 Antibodies

Invitrogen

VEGF Receptor 3 Monoclonal Antibody (AFL4), eBioscience™

Advanced Verification
11 Published Figures
23 References
View all (34) VEGF Receptor 3 antibodies

{{$productOrderCtrl.translations['antibody.pdp.commerceCard.promotion.promotions']}}

{{$productOrderCtrl.translations['antibody.pdp.commerceCard.promotion.viewpromo']}}

{{$productOrderCtrl.translations['antibody.pdp.commerceCard.promotion.promocode']}}: {{promo.promoCode}} {{promo.promoTitle}} {{promo.promoDescription}}. {{$productOrderCtrl.translations['antibody.pdp.commerceCard.promotion.learnmore']}}

Datasheet
Protocols
Questions & Answers
Datasheet
Protocols
Questions & Answers

Cite VEGF Receptor 3 Monoclonal Antibody (AFL4), eBioscience™

  • Published Figures (11)
  • Advanced Verification (1)
VEGF Receptor 3 Antibody in Immunohistochemistry (IHC)
Group 53 Created with Sketch.
VEGF Receptor 3 Antibody in Immunohistochemistry (IHC)
Group 53 Created with Sketch.

FIGURE: 1 / 12

{{ $ctrl.currentElement.advancedVerification.fullName }}
{{ $ctrl.currentElement.advancedVerification.text }}

VEGF Receptor 3 Antibody (14-5988-82) in IHC

Life science alliance 2022 - Figure S5. Increased Vegfr3 expression induced by Tbx1 Cre activation of TgVegfr3 is revealed by qRT-PCR but not by immunofluorescence. (A, B, C, D, E, F, G, H, I, J, K, L) Representative coronal brain sections of Tbx1 Cre/ + (A, B, C, D, E, F) and TgVegfr3; Tbx1 Cre/ + (G, H, I, J, K, L) embryos (E18.5) stained for VEGFR3 (red), VEGFR2 (green), and GFP ( Tbx1 cell fate, white). The cartoon shows the positi... View More {{ $ctrl.currentElement.advancedVerification.fullName }} validation info. View more
Published figure supplied by benchsci-logo
PMID: {{$ctrl.currentElement.benchSciPubmedId}}
View Product

{{$ctrl.videoDesc}}

{{$ctrl.videoLongDesc}}

VEGF Receptor 3 Antibody in Immunohistochemistry (IHC)
VEGF Receptor 3 Antibody in Immunohistochemistry, Immunohistochemistry (Paraffin) (IHC, IHC (P))
VEGF Receptor 3 Antibody in Immunohistochemistry (IHC)
VEGF Receptor 3 Antibody in Immunohistochemistry (IHC)
VEGF Receptor 3 Antibody in Western Blot, Immunohistochemistry (WB, IHC)
VEGF Receptor 3 Antibody in Immunohistochemistry (IHC)
VEGF Receptor 3 Antibody in Western Blot (WB)
VEGF Receptor 3 Antibody in Western Blot (WB)
VEGF Receptor 3 Antibody in Western Blot (WB)
VEGF Receptor 3 Antibody in Flow Cytometry (Flow)
VEGF Receptor 3 Antibody in Flow Cytometry (Flow)
VEGF Receptor 3 Antibody

Product Details

14-5988-82

Applications
Tested Dilution
Publications

Western Blot (WB)

Assay-Dependent
View 7 publications 7 publications

Immunohistochemistry (IHC)

-
View 9 publications 9 publications

Immunohistochemistry (Paraffin) (IHC (P))

-
View 1 publication 1 publication

Immunohistochemistry (Frozen) (IHC (F))

Assay-Dependent
View 1 publication 1 publication

Immunocytochemistry (ICC/IF)

-
View 1 publication 1 publication

Flow Cytometry (Flow)

Assay-Dependent
View 3 publications 3 publications

Immunoprecipitation (IP)

Assay-Dependent
View 1 publication 1 publication

Neutralization (Neu)

Assay-Dependent
-

Functional Assay (Functional)

Assay-Dependent
-
Product Specifications

Species Reactivity

Mouse

Published species

Human, Mouse

Host/Isotype

Rat / IgG2a, kappa

Class

Monoclonal

Type

Antibody

Clone

AFL4

Conjugate

Unconjugated Unconjugated Unconjugated
  • Biotin 
  • Functional Grade
  • Request custom conjugation

Form

Liquid

Concentration

0.5 mg/mL

Purification

Affinity chromatography

Storage buffer

PBS, pH 7.2

Contains

0.09% sodium azide

Storage conditions

4°C

Shipping conditions

Ambient (domestic); Wet ice (international)

RRID

AB_467795

Product Specific Information

Description: The AFL4 monoclonal antibody reacts with the mouse VEGF receptor-3, also known as Flt-4. This 195 kDa molecule was identified as an endothelial-specific member of the receptor tyrosine kinase (RTK) family. During early embryogenesis all endothelial cells express VEGFR-3, while in the adult tissues, VEGFR-3 expression disappears from the vascular endothelial cells and is observed only on the lymphatic endothelium. However, VEGFR-3 expression is induced in the adult tissue upon tumor implementation suggesting an important role for this receptor in the tumor angiogenesis. VEGF-C and VEGF-D bind to and activate VEGFR-3. AFL4 is an antagonist mAb.

Applications Reported: The AFL4 antibody has been reported for use in flow cytometric analysis, immunoprecipitation, immunoblotting (WB), and immunohistochemical staining of frozen tissue sections. It has also been reported in blocking of ligand binding. (Please use Functional Grade purified AFL4, Product # 16-5988, in functional assays).

Applications Tested: The AFL4 antibody has been tested by flow cytometric analysis of in vitro differentiated mouse endothelial cells. In brief, mouse ES cells were incubated on collagen IV matrix for 4 days and subsequently stimulated with VEGF under serum free conditions to induce further differentiation. It is recommended that the antibody be carefully titrated for optimal performance in the assay of interest.

Purity: Greater than 90%, as determined by SDS-PAGE.

Aggregation: Less than 10%, as determined by HPLC.

Filtration: 0.2 µm post-manufacturing filtered.

Target Information

Vascular endothelial growth factor is a key regulator of blood vessel development in embryos and angiogenesis in adult tissues. Unlike VEGF, the related VEGFC stimulates the growth of lymphatic vessels through its specificl ymphatic endothelial receptor VEGFR-3. In 1998, Dumont showed that targeted inactivation of the VEGFR-3 gene in mice resulted in defective blood vessel development in early embryos. Vasculogenesis and angiogenesis occurred, but large vessels became abnormally organized with defective lumens, leading to fluid accumulation in the pericardial cavity and cardiovascular failure at embryonic day 9.5. Thus, VEGFR3 has an essential role in the development of theembryonic cardiovascular system before the emergence of the lymphatic vessels.

For Research Use Only. Not for use in diagnostic procedures. Not for resale without express authorization.

Bioinformatics

Protein Aliases: chylous ascites; FLT-4; Fms-like tyrosine kinase 4; receptor protein tyrosine kinase; soluble VEGFR 3; sVEGFR 3; Tyrosine-protein kinase receptor FLT4; Vascular endothelial growth factor receptor 3; vascular endothelial growth factor receptor-3; VEGF R3; VEGFR-3

View more View less

Gene Aliases: AI323512; Chy; Flt-4; Flt4; VEGFR-3; VEGFR3

View more View less

UniProt ID: (Mouse) P35917

View more View less

Entrez Gene ID: (Mouse) 14257

View more View less

Function(s)
nucleotide binding protein kinase activity protein tyrosine kinase activity transmembrane receptor protein tyrosine kinase activity vascular endothelial growth factor-activated receptor activity protein binding ATP binding kinase activity transferase activity transferase activity, transferring phosphorus-containing groups growth factor binding protein phosphatase binding
Process(es)
angiogenesis positive regulation of protein phosphorylation positive regulation of endothelial cell proliferation vasculature development lymph vessel development lymphangiogenesis sprouting angiogenesis protein phosphorylation transmembrane receptor protein tyrosine kinase signaling pathway positive regulation of cell proliferation positive regulation of vascular endothelial growth factor production positive regulation of endothelial cell migration phosphorylation peptidyl-tyrosine phosphorylation positive regulation of cell growth cellular response to vascular endothelial growth factor stimulus vascular endothelial growth factor signaling pathway negative regulation of apoptotic process positive regulation of MAPK cascade positive regulation of JNK cascade protein autophosphorylation vascular endothelial growth factor receptor signaling pathway blood vessel morphogenesis regulation of blood vessel remodeling positive regulation of ERK1 and ERK2 cascade positive regulation of protein kinase C signaling
It has to be done as per old AB suggested Products section.
guarantee_icon

Performance Guarantee

If an Invitrogen™ antibody doesn't perform as described on our website or datasheet,we'll replace the product at no cost to you, or provide you with a credit for a future purchase.*

Learn more
help_icon

We're here to help

Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.

Contact tech support

Your items have has been added!


Host server : magellan-search-649bfd9c6b-jlsz7:80/100.66.128.142:80.
git-commit: 6001d82975f0049f9efc09ec70b1b2d89229eeb3
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.36.2-2025.09.39-1.0