Disclaimer

Clicking the images or links will redirect you to a website hosted by BenchSci that provides third-party scientific content. Neither the content nor the BenchSci technology and processes for selection have been evaluated by us; we are providing them as-is and without warranty of any kind, including for use or application of the Thermo Fisher Scientific products presented.

  • Primary Antibodies ›
  • TLR6 Antibodies

Abnova

TLR6 Monoclonal Antibody (3D11)

View all (25) TLR6 antibodies

{{$productOrderCtrl.translations['antibody.pdp.commerceCard.promotion.promotions']}}

{{$productOrderCtrl.translations['antibody.pdp.commerceCard.promotion.viewpromo']}}

{{$productOrderCtrl.translations['antibody.pdp.commerceCard.promotion.promocode']}}: {{promo.promoCode}} {{promo.promoTitle}} {{promo.promoDescription}}. {{$productOrderCtrl.translations['antibody.pdp.commerceCard.promotion.learnmore']}}

Datasheet
Protocols
Questions & Answers
Datasheet
Protocols
Questions & Answers

Cite TLR6 Monoclonal Antibody (3D11)

Product Details

H00010333-M09

Applications
Tested Dilution
Publications

ELISA (ELISA)

10 µg/mL
-
Product Specifications

Species Reactivity

Human

Host/Isotype

Mouse / IgG2a, kappa

Class

Monoclonal

Type

Antibody

Clone

3D11

Immunogen

TLR6 (NP_006059, 301 a.a. approximately 430 a.a) full length recombinant protein with GST tag. MW of the GST tag alone is 26 KDa.

Conjugate

Unconjugated Unconjugated Unconjugated

Form

Liquid

Concentration

1 mg/mL

Purification

Protein A

Storage buffer

PBS, pH 7.4

Contains

no preservative

Storage conditions

-20°C, Avoid Freeze/Thaw Cycles

Shipping conditions

Wet ice

Product Specific Information

Immunogen sequence: kttlkaltie hitnqvflf s qtalytvf se mnimmlt isd tpfihm lcph apstf kflnf tqnv ftdsif ekc stlvkle tl ilqknglk d lfkvglmtk dmpsleildv swnslesgr h kenctwve si

Target Information

TLR6 (Toll-like receptor 6) is a member of the toll-like receptor family which plays a fundamental role in pathogen recognition and activation of innate immunity. TLRs are highly conserved and share structural and functional similarities. TLRs recognize pathogen-associated molecular patterns (PAMPs) that are expressed on infectious agents and mediate the production of cytokines necessary for the development of effective immunity. The various TLRs exhibit different patterns of expression. TLR6 functionally interacts with TLR2 to mediate cellular response to bacterial lipoproteins.

For Research Use Only. Not for use in diagnostic procedures. Not for resale without express authorization.

References (0)

Have you cited this product in a publication?
Let us know so we can reference it here.
Cite this product

Bioinformatics

Protein Aliases: CD antigen 286; CD286; Toll-like receptor 6

View more View less

Gene Aliases: CD286; TLR6

View more View less

UniProt ID: (Human) Q9Y2C9

View more View less

Entrez Gene ID: (Human) 10333

View more View less

Function(s)
receptor activity transmembrane signaling receptor activity protein binding Toll-like receptor 2 binding lipopeptide binding
Process(es)
toll-like receptor signaling pathway MyD88-dependent toll-like receptor signaling pathway inflammatory response immune response signal transduction activation of NF-kappaB-inducing kinase activity toll-like receptor 6 signaling pathway toll-like receptor TLR6:TLR2 signaling pathway T-helper 1 type immune response detection of diacyl bacterial lipopeptide defense response to bacterium positive regulation of I-kappaB kinase/NF-kappaB signaling positive regulation of JUN kinase activity innate immune response positive regulation of interleukin-6 biosynthetic process interleukin-1 beta secretion regulation of cytokine secretion cellular response to diacyl bacterial lipopeptide positive regulation of NLRP3 inflammasome complex assembly
It has to be done as per old AB suggested Products section.
guarantee_icon

Performance Guarantee

If an Invitrogen™ antibody doesn't perform as described on our website or datasheet,we'll replace the product at no cost to you, or provide you with a credit for a future purchase.*

Learn more
help_icon

We're here to help

Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.

Contact tech support

Your items have has been added!


Host server : magellan-search-649bfd9c6b-jlsz7:80/100.66.128.142:80.
git-commit: 6001d82975f0049f9efc09ec70b1b2d89229eeb3
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.36.2-2025.09.39-1.0