Disclaimer

Clicking the images or links will redirect you to a website hosted by BenchSci that provides third-party scientific content. Neither the content nor the BenchSci technology and processes for selection have been evaluated by us; we are providing them as-is and without warranty of any kind, including for use or application of the Thermo Fisher Scientific products presented.

  • Primary Antibodies ›
  • TCR gamma/delta Antibodies

Invitrogen

TCR gamma/delta Monoclonal Antibody (eBioGL3 (GL-3, GL3)), FITC, eBioscience™

4 Published Figures
42 References
View all (44) TCR gamma/delta antibodies

{{$productOrderCtrl.translations['antibody.pdp.commerceCard.promotion.promotions']}}

{{$productOrderCtrl.translations['antibody.pdp.commerceCard.promotion.viewpromo']}}

{{$productOrderCtrl.translations['antibody.pdp.commerceCard.promotion.promocode']}}: {{promo.promoCode}} {{promo.promoTitle}} {{promo.promoDescription}}. {{$productOrderCtrl.translations['antibody.pdp.commerceCard.promotion.learnmore']}}

Datasheet
Protocols
Questions & Answers
Datasheet
Protocols
Questions & Answers

Cite TCR gamma/delta Monoclonal Antibody (eBioGL3 (GL-3, GL3)), FITC, eBioscience™

  • Antibody Testing Data (1)
  • Published Figures (4)
TCR gamma/delta Antibody in Flow Cytometry (Flow)
Group 53 Created with Sketch.
TCR gamma/delta Antibody in Flow Cytometry (Flow)
Group 53 Created with Sketch.

FIGURE: 1 / 5

{{ $ctrl.currentElement.advancedVerification.fullName }}
{{ $ctrl.currentElement.advancedVerification.text }}

TCR gamma/delta Antibody (11-5711-82) in Flow

Staining of C57BL/6 lymph node cells (preblocked with Anti-Mouse CD16/32 Purified (Product # 14-0161-82) with Anti-Mouse CD3e APC (Product # 17-0031-82) and 0.25 µg of Armenian Hamster IgG Isotype Control FITC (Product # 11-4888-81) (left) or 0.25 µg of Anti-Mouse gamma delta TCR FITC (right). Cells in the lymphocyte gate were used for analysis. {{ $ctrl.currentElement.advancedVerification.fullName }} validation info. View more
Published figure supplied by benchsci-logo
PMID: {{$ctrl.currentElement.benchSciPubmedId}}
View Product

{{$ctrl.videoDesc}}

{{$ctrl.videoLongDesc}}

TCR gamma/delta Antibody in Flow Cytometry (Flow)
TCR gamma/delta Antibody in Flow Cytometry (Flow)
TCR gamma/delta Antibody in Flow Cytometry (Flow)
TCR gamma/delta Antibody in Flow Cytometry (Flow)
TCR gamma/delta Antibody in Flow Cytometry (Flow)

Product Details

11-5711-82

Applications
Tested Dilution
Publications

Flow Cytometry (Flow)

0.5 µg/test
View 42 publications 42 publications
Product Specifications

Species Reactivity

Mouse

Published species

Bacteria, Mouse

Host/Isotype

Armenian hamster / IgG

Recommended Isotype Control

Armenian Hamster IgG Isotype Control (eBio299Arm), FITC, eBioscience™

Class

Monoclonal

Type

Antibody

Clone

eBioGL3 (GL-3, GL3)

Conjugate

FITC FITC FITC
  • Unconjugated
  • APC 
  • Biotin 
  • Brilliant Violet 650
  • eFluor 450
  • PE 
  • PE-Cyanine5
  • PE-Cyanine7
  • PerCP-eFluor 710
  • Super Bright 645
  • Super Bright 780
  • Request custom conjugation

Excitation/Emission Max

498/517 nm View spectra spectra

Form

Liquid

Concentration

0.5 mg/mL

Purification

Affinity chromatography

Storage buffer

PBS, pH 7.2

Contains

0.09% sodium azide

Storage conditions

4°C, store in dark, DO NOT FREEZE!

Shipping conditions

Ambient (domestic); Wet ice (international)

RRID

AB_465238

Product Specific Information

Description: The GL3 monoclonal antibody (mAb) reacts with the gamma delta T-cell Receptor complex (TCR) on all gamma delta TCR bearing T cells, but it does not react with alpha beta TCR. The gamma delta TCR is present on cells in the thymus, epidermis, epithelial lining of the intestine, peritoneal cavity, and lymphoid tissues.

Applications Reported: This GL3 (GL-3) antibody has been reported for use in flow cytometric analysis.

It is recommended to prestain cells with anti-mouse CD16/32 (Product # 14-0161-85) to prevent non-specific Fc-mediated binding of the GL3 monoclonal antibody.

Applications Tested: This eBioGL3 (GL-3, GL3) antibody has been tested by flow cytometric analysis of mouse lymph node cells. This can be used at less than or equal to 0.5 µg per test. A test is defined as the amount (µg) of antibody that will stain a cell sample in a final volume of 100 µL. Cell number should be determined empirically but can range from 10^5 to 10^8 cells/test. It is recommended that the antibody be carefully titrated for optimal performance in the assay of interest.

Excitation: 488 nm; Emission: 520 nm; Laser: Blue Laser.

Filtration: 0.2 µm post-manufacturing filtered.

Target Information

TCR gamma/delta (T-cell receptor gamma/delta) are specialized T-cells in the immune system. The ability of T cell receptors (TCR) to discriminate foreign from self-peptides presented by major histocompatibility complex (MHC) class II molecules is essential for an effective adaptive immune response. TCR recognition of self-peptides has been linked to autoimmune disease. Mutant self-peptides have been associated with tumors. Engagement of TCRs by a family of bacterial toxins know as superantigens has been responsible for toxic shock syndrome. Autoantibodies to V beta segments of T cell receptors have been isolated from patients with rheumatoid arthritis (RA) and systemic lupus erythematosus (SLE). The autoantibodies block TH1-mediated inflammatory auto-destructive reactions and are believed to be a method by which the immune system compensates for disease. Most human T cells express the TCR alpha-beta and either CD4 or CD8 molecule (single positive, SP). However, a small number of T cells lack both CD4 and CD8 (double negative, DN). Increased percentages of alpha-beta DN T cells have been identified in some autoimmune and immunodeficiency disorders. Gamma-delta T cells are primarily found within the epithelium. They show less TCR diversity and recognize antigens differently than alpha-beta T cells. Subsets of gamma-delta T cells have shown antitumor and immunoregulatory activity.

For Research Use Only. Not for use in diagnostic procedures. Not for resale without express authorization.

How to use the Panel Builder

Watch the video to learn how to use the Invitrogen Flow Cytometry Panel Builder to build your next flow cytometry panel in 5 easy steps.

Bioinformatics

Protein Aliases: T3/TCR complex; TCR delta gamma; TCR gamma/delta; TCR-CD3 complex

View more View less

It has to be done as per old AB suggested Products section.
guarantee_icon

Performance Guarantee

If an Invitrogen™ antibody doesn't perform as described on our website or datasheet,we'll replace the product at no cost to you, or provide you with a credit for a future purchase.*

Learn more
help_icon

We're here to help

Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.

Contact tech support

Your items have has been added!


Host server : magellan-search-585975bbdc-x5cgg:80/100.66.128.25:80.
git-commit: a17be567db00e29ba041ba55be3678c43488a11a
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.37.0-2025.10.41-1.0