Disclaimer

Clicking the images or links will redirect you to a website hosted by BenchSci that provides third-party scientific content. Neither the content nor the BenchSci technology and processes for selection have been evaluated by us; we are providing them as-is and without warranty of any kind, including for use or application of the Thermo Fisher Scientific products presented.

  • Primary Antibodies ›
  • TCR alpha/beta Antibodies

Invitrogen

TCR alpha/beta Monoclonal Antibody (IP26), Biotin, eBioscience™

Advanced Verification
2 Published Figures
2 References
View all (55) TCR alpha/beta antibodies

{{$productOrderCtrl.translations['antibody.pdp.commerceCard.promotion.promotions']}}

{{$productOrderCtrl.translations['antibody.pdp.commerceCard.promotion.viewpromo']}}

{{$productOrderCtrl.translations['antibody.pdp.commerceCard.promotion.promocode']}}: {{promo.promoCode}} {{promo.promoTitle}} {{promo.promoDescription}}. {{$productOrderCtrl.translations['antibody.pdp.commerceCard.promotion.learnmore']}}

Datasheet
Protocols
Questions & Answers
Datasheet
Protocols
Questions & Answers

Cite TCR alpha/beta Monoclonal Antibody (IP26), Biotin, eBioscience™

  • Antibody Testing Data (1)
  • Published Figures (2)
  • Advanced Verification (1)
TCR alpha/beta Antibody in Flow Cytometry (Flow)
Group 53 Created with Sketch.
TCR alpha/beta Antibody in Flow Cytometry (Flow)
Group 53 Created with Sketch.

FIGURE: 1 / 4

{{ $ctrl.currentElement.advancedVerification.fullName }}
{{ $ctrl.currentElement.advancedVerification.text }}

TCR alpha/beta Antibody (13-9986-82) in Flow

Staining of human peripheral blood mononuclear cells with CD45 Pacific Blue, CD3 FITC, CD19 PE and TCR alpha/beta Biotin followed by staining with APC conjugated streptavidin. As expected based on known relative expression patterns, TCR alpha/beta clone IP26 stains a subset of lymphocytes (pink), but not monocytes (orange) and granulocytes (blue). {{ $ctrl.currentElement.advancedVerification.fullName }} validation info. View more
Published figure supplied by benchsci-logo
PMID: {{$ctrl.currentElement.benchSciPubmedId}}
View Product

{{$ctrl.videoDesc}}

{{$ctrl.videoLongDesc}}

TCR alpha/beta Antibody in Flow Cytometry (Flow)
TCR alpha/beta Antibody in Flow Cytometry (Flow)
TCR alpha/beta Antibody in Flow Cytometry (Flow)
TCR alpha/beta Antibody

Product Details

13-9986-82

Applications
Tested Dilution
Publications

Flow Cytometry (Flow)

0.5 µg/test
View 2 publications 2 publications
Product Specifications

Species Reactivity

Human

Host/Isotype

Mouse / IgG1, kappa

Recommended Isotype Control

Mouse IgG1 kappa Isotype Control (P3.6.2.8.1), Biotin, eBioscience™

Class

Monoclonal

Type

Antibody

Clone

IP26

Conjugate

Biotin Biotin Biotin
  • Unconjugated
  • Alexa Fluor 700 
  • APC 
  • APC-eFluor 780
  • eFluor 450
  • FITC
  • NovaFluor Blue 510
  • NovaFluor Blue 610-70S
  • NovaFluor Red 710
  • NovaFluor Yellow 590
  • NovaFluor Yellow 610
  • NovaFluor Yellow 660
  • PE 
  • PE-Cyanine5
  • PE-Cyanine7
  • PE-eFluor 610
  • PerCP-eFluor 710
  • Super Bright 436
  • Super Bright 600
  • Super Bright 780
  • Request custom conjugation

Form

Liquid

Concentration

0.5 mg/mL

Purification

Affinity chromatography

Storage buffer

PBS, pH 7.2

Contains

0.09% sodium azide

Storage conditions

4°C, store in dark, DO NOT FREEZE!

Shipping conditions

Ambient (domestic); Wet ice (international)

RRID

AB_467025

Product Specific Information

Description: The IP26 monoclonal antibody reacts with the alpha beta chain of human TCR.The alpha beta TCR is expressed by the majority of peripheral T cells.

Applications Reported: The IP26 antibody has been reported for use in flow cytometric analysis.

Applications Tested: The IP26 antibody has been tested by flow cytometric analysis of normal human peripheral blood cells. This can be used at less than or equal to 0.5 µg per test. A test is defined as the amount (µg) of antibody that will stain a cell sample in a final volume of 100 µL. Cell number should be determined empirically but can range from 10^5 to 10^8 cells/test. It is recommended that the antibody be carefully titrated for optimal performance in the assay of interest.

Filtration: 0.2 µm post-manufacturing filtered.

Target Information

The T cell antigen receptor (TCR) consists of a ligand-specific alpha/beta heterodimer non-covalently associated with five invariant chains including the CD3 gamma/delta/eta and zeta subunits, all of which are required for efficient surface expression. T cell activation through the TCR induces cellular differentiation and/or proliferation and the production of lymphokines and cytokines. Both the CD3 and TCR zeta subunits are proposed to be responsible for the intracellular signal transduction events. Majority of T cells present in the blood, lymph and secondary lymphoid organs express TCR alpha/beta heterodimers, whereas the T cells expressing TCR gamma/delta heterodimers are localized mainly in epithelial tissues and at the sites of infection. The subunits of TCR heterodimers are covalently bonded and associate with the CD3 subunits in the endoplasmic reticulum to form functional TCR-CD3 complex. Lack of expression of any of the chains is sufficient to stop cell surface expression. The ability of T cell receptors (TCR) to discriminate foreign from self-peptides presented by major histocompatibility complex (MHC) class II molecules is essential for an effective adaptive immune response. TCR recognition of self-peptides has been linked to autoimmune disease. Mutant self-peptides have been associated with tumors. Engagement of TCRs by a family of bacterial toxins know as superantigens has been responsible for toxic shock syndrome. Autoantibodies to V beta segments of T cell receptors have been isolated from patients with rheumatoid arthritis (RA) and systemic lupus erythematosus (SLE). The autoantibodies block TH1-mediated inflammatory auto-destructive reactions and are believed to be a method by which the immune system compensates for disease (ref5). T Cell and TCR Diversity Most human T cells express the TCR alpha-beta and either CD4 or CD8 molecule (single positive, SP). A small number of T cells lack both CD4 and CD8 (double negative, DN). Increased percentages of alpha-beta DN T cells have been identified in some autoimmune and immunodeficiency disorders. Gamma-delta T cells are primarily found within the epithelium. They show less TCR diversity and recognize antigens differently than alpha-beta T cells. Subsets of gamma-delta T cells have shown antitumor and immunoregulatory activity.

For Research Use Only. Not for use in diagnostic procedures. Not for resale without express authorization.

Bioinformatics

Protein Aliases: FLJ22602; MGC117436; MGC22624; MGC23964; MGC71411; t-cell antigen receptor; T3/TCR complex; tcr alpha; TCR alpha/ beta; TCR beta

View more View less

Gene Aliases: IMD7; TCRA; TCRB; TRA; TRA@; TRB@; TRCA

View more View less

Entrez Gene ID: (Human) 6955, (Human) 28755, (Human) 6957

View more View less

It has to be done as per old AB suggested Products section.
guarantee_icon

Performance Guarantee

If an Invitrogen™ antibody doesn't perform as described on our website or datasheet,we'll replace the product at no cost to you, or provide you with a credit for a future purchase.*

Learn more
help_icon

We're here to help

Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.

Contact tech support

Your items have has been added!


Host server : magellan-search-649bfd9c6b-jlsz7:80/100.66.128.142:80.
git-commit: 6001d82975f0049f9efc09ec70b1b2d89229eeb3
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.36.2-2025.09.39-1.0