Disclaimer

Clicking the images or links will redirect you to a website hosted by BenchSci that provides third-party scientific content. Neither the content nor the BenchSci technology and processes for selection have been evaluated by us; we are providing them as-is and without warranty of any kind, including for use or application of the Thermo Fisher Scientific products presented.

  • Primary Antibodies ›
  • TBCEL Antibodies

OriGene

TBCEL Monoclonal Antibody (OTI2A9), TrueMAB™

View all (10) TBCEL antibodies

{{$productOrderCtrl.translations['antibody.pdp.commerceCard.promotion.promotions']}}

{{$productOrderCtrl.translations['antibody.pdp.commerceCard.promotion.viewpromo']}}

{{$productOrderCtrl.translations['antibody.pdp.commerceCard.promotion.promocode']}}: {{promo.promoCode}} {{promo.promoTitle}} {{promo.promoDescription}}. {{$productOrderCtrl.translations['antibody.pdp.commerceCard.promotion.learnmore']}}

Datasheet
Protocols
Questions & Answers
Datasheet
Protocols
Questions & Answers

Cite TBCEL Monoclonal Antibody (OTI2A9), TrueMAB™

  • Antibody Testing Data (5)
TBCEL Antibody in Immunohistochemistry (Paraffin) (IHC (P))
Group 53 Created with Sketch.
TBCEL Antibody in Immunohistochemistry (Paraffin) (IHC (P))
Group 53 Created with Sketch.

FIGURE: 1 / 5

{{ $ctrl.currentElement.advancedVerification.fullName }}
{{ $ctrl.currentElement.advancedVerification.text }}

TBCEL Antibody (CF502672) in IHC (P)

Immunohistochemical staining of paraffin-embedded human lymph node tissue within the normal limits using anti-TBCEL mouse monoclonal antibody. (Heat-induced epitope retrieval by 10mM citric buffer, pH6.0, 100°C for 10min, TA502672) {{ $ctrl.currentElement.advancedVerification.fullName }} validation info. View more
Published figure supplied by benchsci-logo
PMID: {{$ctrl.currentElement.benchSciPubmedId}}
View Product

{{$ctrl.videoDesc}}

{{$ctrl.videoLongDesc}}

TBCEL Antibody in Immunohistochemistry (Paraffin) (IHC (P))
TBCEL Antibody in Immunohistochemistry (Paraffin) (IHC (P))
TBCEL Antibody in Immunohistochemistry (Paraffin) (IHC (P))
TBCEL Antibody in Western Blot (WB)
TBCEL Antibody in Western Blot (WB)
TBCEL Monoclonal Antibody (OTI2A9), TrueMAB™

Product Details

CF502672

Applications
Tested Dilution
Publications

Western Blot (WB)

1:2,000
-

Immunohistochemistry (Paraffin) (IHC (P))

1:150
-
Product Specifications

Species Reactivity

Human

Host/Isotype

Mouse / IgG2a

Class

Monoclonal

Type

Antibody

Clone

OTI2A9

Immunogen

Full length human recombinant protein of human TBCEL produced in HEK293T cell.
View immunogen

Conjugate

Unconjugated Unconjugated Unconjugated

Form

Lyophilized

Concentration

1 mg/mL

Purification

Affinity chromatography

Storage buffer

PBS, pH 7.3, with 8% trehalose

Contains

no preservative

Storage conditions

-20°C, Avoid Freeze/Thaw Cycles

Shipping conditions

Ambient (domestic); Wet ice (international)

Product Specific Information

For reconstitution, we recommend adding 100 µL distilled water to a final antibody concentration of about 1 mg/mL. To use this carrier-free antibody for conjugation experiments, we strongly recommend performing another round of desalting. (Zeba Spin Desalting Columns, 7KMWCO, 0.5 mL, Product # 89882)

Target Information

Acts as a regulator of tubulin stability.

For Research Use Only. Not for use in diagnostic procedures. Not for resale without express authorization.

References (0)

Have you cited this product in a publication?
Let us know so we can reference it here.
Cite this product

Bioinformatics

Protein Aliases: catastrophin; E-like; EL; leucine rich repeat containing 35; Leucine-rich repeat-containing protein 35; tubulin folding cofactor E-like; Tubulin-specific chaperone cofactor E-like protein; tubulin-specific chaperone e-like

View more View less

Gene Aliases: El; LRRC35; TBCEL

View more View less

UniProt ID: (Human) Q5QJ74

View more View less

Entrez Gene ID: (Human) 219899

View more View less

Function(s)
protein binding
It has to be done as per old AB suggested Products section.
guarantee_icon

Performance Guarantee

If an Invitrogen™ antibody doesn't perform as described on our website or datasheet,we'll replace the product at no cost to you, or provide you with a credit for a future purchase.*

Learn more
help_icon

We're here to help

Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.

Contact tech support

Your items have has been added!


Host server : magellan-search-649bfd9c6b-jlsz7:80/100.66.128.142:80.
git-commit: 6001d82975f0049f9efc09ec70b1b2d89229eeb3
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.36.2-2025.09.39-1.0