Disclaimer

Clicking the images or links will redirect you to a website hosted by BenchSci that provides third-party scientific content. Neither the content nor the BenchSci technology and processes for selection have been evaluated by us; we are providing them as-is and without warranty of any kind, including for use or application of the Thermo Fisher Scientific products presented.

  • Primary Antibodies ›
  • SPTBN2 Antibodies

NeoBiotechnologies

Spectrin beta III (SPTBN2) Monoclonal Antibody (SPTBN2/1778)

View all (29) SPTBN2 antibodies

{{$productOrderCtrl.translations['antibody.pdp.commerceCard.promotion.promotions']}}

{{$productOrderCtrl.translations['antibody.pdp.commerceCard.promotion.viewpromo']}}

{{$productOrderCtrl.translations['antibody.pdp.commerceCard.promotion.promocode']}}: {{promo.promoCode}} {{promo.promoTitle}} {{promo.promoDescription}}. {{$productOrderCtrl.translations['antibody.pdp.commerceCard.promotion.learnmore']}}

Datasheet
Protocols
Questions & Answers
Datasheet
Protocols
Questions & Answers

Cite Spectrin beta III (SPTBN2) Monoclonal Antibody (SPTBN2/1778)

  • Antibody Testing Data (6)
Spectrin beta III (SPTBN2) Antibody in Immunocytochemistry (ICC/IF)
Group 53 Created with Sketch.
Spectrin beta III (SPTBN2) Antibody in Immunocytochemistry (ICC/IF)
Group 53 Created with Sketch.

FIGURE: 1 / 6

{{ $ctrl.currentElement.advancedVerification.fullName }}
{{ $ctrl.currentElement.advancedVerification.text }}

Spectrin beta III (SPTBN2) Antibody (6712-MSM5-P0) in ICC/IF

Immunofluorescence analysis of MeOH-fixed HeLa cells labeling Spectrin beta III with Spectrin beta III Mouse Monoclonal Antibody (SPTBN2/1778) followed by goat anti-mouse IgG-CF488 (green). Nuclei stained with RedDot (red). {{ $ctrl.currentElement.advancedVerification.fullName }} validation info. View more
Published figure supplied by benchsci-logo
PMID: {{$ctrl.currentElement.benchSciPubmedId}}
View Product

{{$ctrl.videoDesc}}

{{$ctrl.videoLongDesc}}

Spectrin beta III (SPTBN2) Antibody in Immunocytochemistry (ICC/IF)
Spectrin beta III (SPTBN2) Antibody in Immunohistochemistry (Paraffin) (IHC (P))
Spectrin beta III (SPTBN2) Antibody in Western Blot (WB)
Spectrin beta III (SPTBN2) Antibody in Western Blot (WB)
Spectrin beta III (SPTBN2) Antibody in Peptide array (ARRAY)
Spectrin beta III (SPTBN2) Antibody in SDS-PAGE (SDS-PAGE)
Spectrin beta III (SPTBN2) Monoclonal Antibody (SPTBN2/1778)

Product Details

6712-MSM5-P0

Applications
Tested Dilution
Publications

Western Blot (WB)

1-2 µg/mL
-

Immunohistochemistry (Paraffin) (IHC (P))

Assay-dependent
-

Immunohistochemistry (PFA fixed) (IHC (PFA))

1-2 µg/mL
-

Immunocytochemistry (ICC/IF)

2-4 µg/mL
-

Peptide Array (Array)

Assay-dependent
-
Product Specifications

Species Reactivity

Human

Host/Isotype

Mouse / IgG1, kappa

Class

Monoclonal

Type

Antibody

Clone

SPTBN2/1778

Immunogen

Recombinant fragment (around aa356-475) of human SPTBN2 protein.
View immunogen

Conjugate

Unconjugated Unconjugated Unconjugated

Form

Liquid

Concentration

200 µg/mL

Purification

Protein A/G

Storage buffer

PBS, pH 7.4, with 0.05% BSA

Contains

0.05% sodium azide

Storage conditions

4°C, do not freeze

Shipping conditions

Wet ice

Target Information

Spectrins are principle components of a cell's membrane-cytoskeleton and are composed of two alpha and two beta spectrin subunits. The protein encoded by this gene (SPTBN2), is called spectrin beta non-erythrocytic 2 or beta-III spectrin. It is related to, but distinct from, the beta-II spectrin gene which is also known as spectrin beta non-erythrocytic 1 (SPTBN1). SPTBN2 regulates the glutamate signaling pathway by stabilizing the glutamate transporter EAAT4 at the surface of the plasma membrane. Mutations in this gene cause a form of spinocerebellar ataxia, SCA5, that is characterized by neurodegeneration, progressive locomotor incoordination, dysarthria, and uncoordinated eye movements.

For Research Use Only. Not for use in diagnostic procedures. Not for resale without express authorization.

References (0)

Have you cited this product in a publication?
Let us know so we can reference it here.
Cite this product

Bioinformatics

Protein Aliases: Beta-III spectrin; glutamate transporter EAAT4-associated protein 41; Spectrin Beta; spectrin beta chain, brain 2; Spectrin beta chain, non-erythrocytic 2; spectrin beta III sigma 2; spectrin, beta, non-erythrocytic 2; spectrin, non-erythroid beta chain 2; Spinocerebellar ataxia 5 protein

View more View less

Gene Aliases: GTRAP41; KIAA0302; SCA5; SCAR14; SPTBN2

View more View less

UniProt ID: (Human) O15020

View more View less

Entrez Gene ID: (Human) 6712

View more View less

Function(s)
actin binding Ras guanyl-nucleotide exchange factor activity structural constituent of cytoskeleton phospholipid binding cadherin binding involved in cell-cell adhesion
Process(es)
MAPK cascade ER to Golgi vesicle-mediated transport cytoskeleton organization axon guidance synapse assembly vesicle-mediated transport antigen processing and presentation of exogenous peptide antigen via MHC class II cerebellar Purkinje cell layer morphogenesis adult behavior multicellular organism growth positive regulation of GTPase activity actin filament capping cellular response to ethanol cell-cell adhesion
It has to be done as per old AB suggested Products section.
guarantee_icon

Performance Guarantee

If an Invitrogen™ antibody doesn't perform as described on our website or datasheet,we'll replace the product at no cost to you, or provide you with a credit for a future purchase.*

Learn more
help_icon

We're here to help

Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.

Contact tech support

Your items have has been added!


Host server : magellan-search-5fc94fc45-hqqk4:80/100.66.128.142:80.
git-commit: 7a6627b71d15189168043ee5c63248cc100e5965
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.37.0-2025.10.41-1.0