Disclaimer

Clicking the images or links will redirect you to a website hosted by BenchSci that provides third-party scientific content. Neither the content nor the BenchSci technology and processes for selection have been evaluated by us; we are providing them as-is and without warranty of any kind, including for use or application of the Thermo Fisher Scientific products presented.

  • Primary Antibodies ›
  • SRF Antibodies

NeoBiotechnologies

Serum Response Element Binding Transcription Factor (SRF) Monoclonal Antibody (PCRP-SRF-1F7)

View all (31) SRF antibodies

{{$productOrderCtrl.translations['antibody.pdp.commerceCard.promotion.promotions']}}

{{$productOrderCtrl.translations['antibody.pdp.commerceCard.promotion.viewpromo']}}

{{$productOrderCtrl.translations['antibody.pdp.commerceCard.promotion.promocode']}}: {{promo.promoCode}} {{promo.promoTitle}} {{promo.promoDescription}}. {{$productOrderCtrl.translations['antibody.pdp.commerceCard.promotion.learnmore']}}

Datasheet
Protocols
Questions & Answers
Datasheet
Protocols
Questions & Answers

Cite Serum Response Element Binding Transcription Factor (SRF) Monoclonal Antibody (PCRP-SRF-1F7)

  • Antibody Testing Data (5)
Serum Response Element Binding Transcription Factor (SRF) Antibody in Immunocytochemistry (ICC/IF)
Group 53 Created with Sketch.
Serum Response Element Binding Transcription Factor (SRF) Antibody in Immunocytochemistry (ICC/IF)
Group 53 Created with Sketch.

FIGURE: 1 / 5

{{ $ctrl.currentElement.advancedVerification.fullName }}
{{ $ctrl.currentElement.advancedVerification.text }}

Serum Response Element Binding Transcription Factor (SRF) Antibody (6722-MSM2-P0) in ICC/IF

Immunofluorescence analysis of PFA-fixed HeLa cells stained using SRF Mouse Monoclonal Antibody (PCRP-SRF-1F7) followed by goat anti-mouse IgG-CF488 (green). CF640R phalloidin (red). {{ $ctrl.currentElement.advancedVerification.fullName }} validation info. View more
Published figure supplied by benchsci-logo
PMID: {{$ctrl.currentElement.benchSciPubmedId}}
View Product

{{$ctrl.videoDesc}}

{{$ctrl.videoLongDesc}}

Serum Response Element Binding Transcription Factor (SRF) Antibody in Immunocytochemistry (ICC/IF)
Serum Response Element Binding Transcription Factor (SRF) Antibody in Immunohistochemistry (Paraffin) (IHC (P))
Serum Response Element Binding Transcription Factor (SRF) Antibody in Flow Cytometry (Flow)
Serum Response Element Binding Transcription Factor (SRF) Antibody in Peptide array (ARRAY)
Serum Response Element Binding Transcription Factor (SRF) Antibody in SDS-PAGE (SDS-PAGE)
Serum Response Element Binding Transcription Factor (SRF) Monoclonal Antibody (PCRP-SRF-1F7)

Product Details

6722-MSM2-P0

Applications
Tested Dilution
Publications

Western Blot (WB)

1-2 µg/mL
-

Immunohistochemistry (Paraffin) (IHC (P))

2 µg/mL
-

Immunohistochemistry (PFA fixed) (IHC (PFA))

1-2 µg/mL
-

Immunocytochemistry (ICC/IF)

1-2 µg/mL
-

Flow Cytometry (Flow)

1-2 µg/10^6 cells
-

ELISA (ELISA)

Assay-dependent
-

Immunoprecipitation (IP)

1-2 µg
-

Peptide Array (Array)

Assay-dependent
-
Product Specifications

Species Reactivity

Human

Host/Isotype

Mouse / IgG2b

Class

Monoclonal

Type

Antibody

Clone

PCRP-SRF-1F7

Immunogen

Recombinant full-length human SRF protein
View immunogen

Conjugate

Unconjugated Unconjugated Unconjugated

Form

Liquid

Concentration

200 µg/mL

Purification

Protein A/G

Storage buffer

PBS, pH 7.4, with 0.05% BSA

Contains

0.05% sodium azide

Storage conditions

4°C, do not freeze

Shipping conditions

Wet ice

Target Information

Serum response factor (SRF) is a transcription factor that binds the serum response element (SRE), a sequence that mediates the transient response of many cellular genes to growth stimulation. SRF-binding sites are also constitutive promotor elements in many muscle-specific promotors. SFR is required for cardiac differentiation and maturation.

For Research Use Only. Not for use in diagnostic procedures. Not for resale without express authorization.

References (0)

Have you cited this product in a publication?
Let us know so we can reference it here.
Cite this product

Bioinformatics

Protein Aliases: c-fos serum response element-binding transcription factor; minichromosome maintenance 1 homolog; OTTHUMP00000039820; Serum response factor; serum response factor (c-fos serum response element-binding transcription factor); SRF

View more View less

Gene Aliases: MCM1; SRF

View more View less

UniProt ID: (Human) P11831

View more View less

Entrez Gene ID: (Human) 6722

View more View less

Function(s)
RNA polymerase II core promoter proximal region sequence-specific DNA binding transcription factor activity, RNA polymerase II core promoter sequence-specific transcription factor activity, RNA polymerase II transcription factor binding transcriptional activator activity, RNA polymerase II core promoter proximal region sequence-specific binding transcriptional activator activity, RNA polymerase II transcription regulatory region sequence-specific binding transcription factor activity, sequence-specific DNA binding transcription factor activity, RNA polymerase II distal enhancer sequence-specific binding protein binding transcription factor binding serum response element binding chromatin DNA binding protein homodimerization activity histone deacetylase binding primary miRNA binding
Process(es)
patterning of blood vessels response to hypoxia mesoderm formation neuron migration trophectodermal cell differentiation heart looping morphogenesis of an epithelial sheet cell migration involved in sprouting angiogenesis positive regulation of transcription from RNA polymerase II promoter involved in myocardial precursor cell differentiation transcription from RNA polymerase II promoter cell-matrix adhesion heart development long-term memory negative regulation of cell proliferation associative learning response to toxic substance response to hormone epithelial structure maintenance positive regulation of transcription via serum response element binding hippocampus development tangential migration from the subventricular zone to the olfactory bulb regulation of cell adhesion platelet activation platelet formation negative regulation of cell migration thyroid gland development neuron projection development regulation of water loss via skin response to cytokine megakaryocyte development dorsal aorta morphogenesis mRNA transcription from RNA polymerase II promoter stress fiber assembly skin morphogenesis positive thymic T cell selection sarcomere organization positive regulation of cell differentiation positive regulation of axon extension positive regulation of transcription from RNA polymerase II promoter positive regulation of smooth muscle contraction positive regulation of transcription by glucose muscle cell cellular homeostasis thymus development developmental growth neuron development erythrocyte development positive regulation of sequence-specific DNA binding transcription factor activity regulation of smooth muscle cell differentiation positive regulation of filopodium assembly cardiac myofibril assembly angiogenesis involved in wound healing hematopoietic stem cell differentiation positive regulation of transcription initiation from RNA polymerase II promoter long term synaptic depression face development heart trabecula formation lung morphogenesis bronchus cartilage development trachea cartilage development cardiac vascular smooth muscle cell differentiation eyelid development in camera-type eye lung smooth muscle development bicellular tight junction assembly cellular response to glucose stimulus primitive streak formation epithelial cell-cell adhesion cellular senescence negative regulation of beta-amyloid clearance negative regulation of pri-miRNA transcription from RNA polymerase II promoter positive regulation of pri-miRNA transcription from RNA polymerase II promoter
It has to be done as per old AB suggested Products section.
guarantee_icon

Performance Guarantee

If an Invitrogen™ antibody doesn't perform as described on our website or datasheet,we'll replace the product at no cost to you, or provide you with a credit for a future purchase.*

Learn more
help_icon

We're here to help

Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.

Contact tech support

Your items have has been added!


Host server : magellan-search-585975bbdc-x5cgg:80/100.66.128.25:80.
git-commit: a17be567db00e29ba041ba55be3678c43488a11a
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.37.0-2025.10.41-1.0