Disclaimer

Clicking the images or links will redirect you to a website hosted by BenchSci that provides third-party scientific content. Neither the content nor the BenchSci technology and processes for selection have been evaluated by us; we are providing them as-is and without warranty of any kind, including for use or application of the Thermo Fisher Scientific products presented.

  • Primary Antibodies ›
  • Unc18-2 Antibodies

Proteintech

STXBP2 Polyclonal Antibody

5 References
View all (12) Unc18-2 antibodies

{{$productOrderCtrl.translations['antibody.pdp.commerceCard.promotion.promotions']}}

{{$productOrderCtrl.translations['antibody.pdp.commerceCard.promotion.viewpromo']}}

{{$productOrderCtrl.translations['antibody.pdp.commerceCard.promotion.promocode']}}: {{promo.promoCode}} {{promo.promoTitle}} {{promo.promoDescription}}. {{$productOrderCtrl.translations['antibody.pdp.commerceCard.promotion.learnmore']}}

Datasheet
Protocols
Questions & Answers
Datasheet
Protocols
Questions & Answers

Cite STXBP2 Polyclonal Antibody

  • Antibody Testing Data (9)
STXBP2 Antibody in Immunohistochemistry (Paraffin) (IHC (P))
Group 53 Created with Sketch.
STXBP2 Antibody in Immunohistochemistry (Paraffin) (IHC (P))
Group 53 Created with Sketch.

FIGURE: 1 / 9

{{ $ctrl.currentElement.advancedVerification.fullName }}
{{ $ctrl.currentElement.advancedVerification.text }}

STXBP2 Antibody (15312-1-AP) in IHC (P)

Immunohistochemistry of paraffin-embedded human lung cancer using 15312-1-AP (STXBP2 antibody) at dilution of 1:50 (under 40x lens). {{ $ctrl.currentElement.advancedVerification.fullName }} validation info. View more
Published figure supplied by benchsci-logo
PMID: {{$ctrl.currentElement.benchSciPubmedId}}
View Product

{{$ctrl.videoDesc}}

{{$ctrl.videoLongDesc}}

STXBP2 Antibody in Immunohistochemistry (Paraffin) (IHC (P))
STXBP2 Antibody in Immunohistochemistry (Paraffin) (IHC (P))
STXBP2 Antibody in Immunohistochemistry (Paraffin) (IHC (P))
STXBP2 Antibody in Immunohistochemistry (Paraffin) (IHC (P))
STXBP2 Antibody in Western Blot (WB)
STXBP2 Antibody in Western Blot (WB)
STXBP2 Antibody in Western Blot (WB)
STXBP2 Antibody in Western Blot (WB)
STXBP2 Antibody in Western Blot (WB)
STXBP2 Polyclonal Antibody

Product Details

15312-1-AP

Applications
Tested Dilution
Publications

Western Blot (WB)

1:500-1:2,000
View 5 publications 5 publications

Immunohistochemistry (Paraffin) (IHC (P))

1:50-1:200
-
Product Specifications

Species Reactivity

Human, Mouse

Published species

Human

Host/Isotype

Rabbit / IgG

Class

Polyclonal

Type

Antibody

Immunogen

STXBP2 Fusion Protein Ag7537 (241-590 aa encoded by BC002869)
View immunogen

Conjugate

Unconjugated Unconjugated Unconjugated

Form

Liquid

Concentration

0.21 mg/mL

Purification

Antigen affinity chromatography

Storage buffer

PBS, pH 7.3, with 50% glycerol

Contains

0.02% sodium azide

Storage conditions

-20°C

Shipping conditions

Wet ice (domestic); Dry ice (international)

Product Specific Information

Immunogen sequence: LHELTFQAM AYDLLDIEQD TYRYETTGLS EAREKAVLLD EDDDLWVELR HMHIADVSKK VTELLRTFCE SKRLTTDKAN IKDLSQILKK MPQYQKELNK YSTHLHLADD CMKHFKGSVE KLCSVEQDLA MGSDAEGEKI KDSMKLIVPV LLDAAVPAYD KIRVLLLYIL LRNGVSEENL AKLIQHANVQ AHSSLIRNLE QLGGTVTNPG GSGTSSRLEP RERMEPTYQL SRWTPVIKDV MEDAVEDRLD RNLWPFVSDP APTASSQAAV SARFGHWHKN KAGIEARAGP RLIVYVMGGV AMSEMRAAYE VTRATEGKWE VLIGSSHILT PTRFLDDLKA LDKKLEDIAL P (241-590 aa encoded by BC002869)

Target Information

This gene encodes a member of the STXBP/unc-18/SEC1 family. The encoded protein is involved in intracellular trafficking, control of SNARE (soluble NSF attachment protein receptor) complex assembly, and the release of cytotoxic granules by natural killer cells. Mutations in this gene are associated with familial hemophagocytic lymphohistiocytosis. Alternatively spliced transcript variants encoding different isoforms have been noted for this gene.

For Research Use Only. Not for use in diagnostic procedures. Not for resale without express authorization.

Bioinformatics

Protein Aliases: Munc18-2; MUSEC1; Protein unc-18 homolog 2; Protein unc-18 homolog B; syntaxin binding protein Munc18-2; Syntaxin-binding protein 2; unc-18 homolog 2; Unc-18B; Unc18-2

View more View less

Gene Aliases: C79054; FHL5; Hunc18b; Munc-18-2; Munc-18b; MUNC18-2; Munc18b; muSec1; pp10122; STXBP2; Sxtbp2; Sxtp2; UNC18-2; UNC18B

View more View less

UniProt ID: (Human) Q15833, (Mouse) Q64324

View more View less

Entrez Gene ID: (Human) 6813, (Mouse) 20911

View more View less

Function(s)
protein binding syntaxin-1 binding syntaxin-3 binding syntaxin binding
Process(es)
leukocyte mediated cytotoxicity transport exocytosis vesicle docking involved in exocytosis protein transport vesicle-mediated transport regulation of mast cell degranulation neutrophil degranulation
It has to be done as per old AB suggested Products section.
guarantee_icon

Performance Guarantee

If an Invitrogen™ antibody doesn't perform as described on our website or datasheet,we'll replace the product at no cost to you, or provide you with a credit for a future purchase.*

Learn more
help_icon

We're here to help

Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.

Contact tech support

Your items have has been added!


Host server : magellan-search-649bfd9c6b-jlsz7:80/100.66.128.142:80.
git-commit: 6001d82975f0049f9efc09ec70b1b2d89229eeb3
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.36.2-2025.09.39-1.0