Disclaimer

Clicking the images or links will redirect you to a website hosted by BenchSci that provides third-party scientific content. Neither the content nor the BenchSci technology and processes for selection have been evaluated by us; we are providing them as-is and without warranty of any kind, including for use or application of the Thermo Fisher Scientific products presented.

  • Primary Antibodies ›
  • SOX2 Antibodies

NeoBiotechnologies

SOX2 (Embryonic Stem Cell Marker) Monoclonal Antibody (rSOX2/1791)

View all (88) SOX2 antibodies

{{$productOrderCtrl.translations['antibody.pdp.commerceCard.promotion.promotions']}}

{{$productOrderCtrl.translations['antibody.pdp.commerceCard.promotion.viewpromo']}}

{{$productOrderCtrl.translations['antibody.pdp.commerceCard.promotion.promocode']}}: {{promo.promoCode}} {{promo.promoTitle}} {{promo.promoDescription}}. {{$productOrderCtrl.translations['antibody.pdp.commerceCard.promotion.learnmore']}}

Datasheet
Protocols
Questions & Answers
Datasheet
Protocols
Questions & Answers

Cite SOX2 (Embryonic Stem Cell Marker) Monoclonal Antibody (rSOX2/1791)

SOX2 (Embryonic Stem Cell Marker) Monoclonal Antibody (rSOX2/1791)
  • Antibody Testing Data (2)
SOX2 (Embryonic Stem Cell Marker) Antibody in Immunohistochemistry (Paraffin) (IHC (P))
Group 53 Created with Sketch.
SOX2 (Embryonic Stem Cell Marker) Antibody in Immunohistochemistry (Paraffin) (IHC (P))
Group 53 Created with Sketch.

FIGURE: 1 / 2

{{ $ctrl.currentElement.advancedVerification.fullName }}
{{ $ctrl.currentElement.advancedVerification.text }}

SOX2 (Embryonic Stem Cell Marker) Antibody (6657-MSM4-P0) in IHC (P)

Formalin-fixed, paraffin-embedded human cervix stained with SOX2 Recombinant Mouse Monoclonal Antibody (rSOX2/1791). {{ $ctrl.currentElement.advancedVerification.fullName }} validation info. View more
Published figure supplied by benchsci-logo
PMID: {{$ctrl.currentElement.benchSciPubmedId}}
View Product

{{$ctrl.videoDesc}}

{{$ctrl.videoLongDesc}}

SOX2 (Embryonic Stem Cell Marker) Antibody in Immunohistochemistry (Paraffin) (IHC (P))
SOX2 (Embryonic Stem Cell Marker) Antibody in SDS-PAGE (SDS-PAGE)

Product Details

6657-MSM4-P0

Applications
Tested Dilution
Publications

Immunohistochemistry (Paraffin) (IHC (P))

Assay-dependent
-

Immunohistochemistry (PFA fixed) (IHC (PFA))

1-2 µg/mL
-
Product Specifications

Species Reactivity

Human, Mouse

Host/Isotype

Mouse / IgG1, kappa

Class

Monoclonal

Type

Antibody

Clone

rSOX2/1791

Immunogen

Recombinant fragment (within aa176-305) of human SOX2 protein.
View immunogen

Conjugate

Unconjugated Unconjugated Unconjugated

Form

Liquid

Concentration

200 µg/mL

Purification

Protein A/G

Storage buffer

PBS, pH 7.4, with 0.05% BSA

Contains

0.05% sodium azide

Storage conditions

4°C, do not freeze

Shipping conditions

Wet ice

Target Information

SOX2 is an intronless gene encoding a member of the SRY-related HMG-box (SOX) family of transcription factors involved in the regulation of embryonic development and in the determination of cell fate. The product of the SOX2 gene is required for stem-cell maintenance in the central nervous system, and also regulates gene expression in the stomach. The SOX2 gene lies within an intron of another gene called SOX2 overlapping transcript (SOX2OT). Further, SOX2 protein may act as a transcriptional activator after forming a protein complex with other proteins. Mutations in the SOX2 gene have been associated with bilateral anophthalmia, a severe form of structural eye malformation, optic nerve hypoplasia and syndromic microphthalmia.

For Research Use Only. Not for use in diagnostic procedures. Not for resale without express authorization.

References (0)

Have you cited this product in a publication?
Let us know so we can reference it here.
Cite this product

Bioinformatics

Protein Aliases: Delta EF2a; MCOPS3 (Microphthalmia Syndromic type 3); sex determining region Y-box 2; SOX 2; SRY (sex determining region Y) box 2; SRY (sex determining region Y)-box 2; SRY box 2; SRY related HMG box 2; SRY-box containing gene 2; SRY-related HMG-box gene 2; Transcription factor SOX-2; transcription factor SOX2

View more View less

Gene Aliases: ANOP3; lcc; MCOPS3; Sox-2; SOX2; ysb

View more View less

UniProt ID: (Human) P48431, (Mouse) P48432

View more View less

Entrez Gene ID: (Human) 6657, (Mouse) 20674

View more View less

Function(s)
transcription regulatory region sequence-specific DNA binding transcriptional activator activity, RNA polymerase II core promoter proximal region sequence-specific binding DNA binding transcription factor activity, sequence-specific DNA binding protein binding miRNA binding sequence-specific DNA binding transcription regulatory region DNA binding RNA polymerase II transcription factor activity, sequence-specific DNA binding chromatin binding transcription factor binding chromatin DNA binding protein heterodimerization activity
Process(es)
negative regulation of transcription from RNA polymerase II promoter osteoblast differentiation eye development endodermal cell fate specification chromatin organization regulation of transcription, DNA-templated transcription from RNA polymerase II promoter cell cycle arrest response to wounding regulation of gene expression glial cell fate commitment pituitary gland development adenohypophysis development positive regulation of cell-cell adhesion forebrain development somatic stem cell population maintenance tissue regeneration regulation of cysteine-type endopeptidase activity involved in apoptotic process positive regulation of MAPK cascade positive regulation of cell differentiation negative regulation of neuron differentiation positive regulation of transcription, DNA-templated positive regulation of transcription from RNA polymerase II promoter inner ear development negative regulation of epithelial cell proliferation response to growth factor negative regulation of canonical Wnt signaling pathway neuronal stem cell population maintenance cell fate specification positive regulation of neuroblast proliferation transcription, DNA-templated regulation of transcription from RNA polymerase II promoter multicellular organism development sensory perception of sound response to organic substance stem cell population maintenance forebrain neuron differentiation cerebral cortex development negative regulation of Wnt signaling pathway male genitalia development positive regulation of epithelial cell differentiation olfactory placode formation response to retinoic acid inner ear morphogenesis tongue development cell fate commitment negative regulation of cell differentiation positive regulation of neuron differentiation negative regulation of osteoblast differentiation positive regulation of Notch signaling pathway pigment biosynthetic process lung alveolus development embryonic organ development anatomical structure formation involved in morphogenesis neuron fate commitment diencephalon morphogenesis stem cell differentiation regulation of neurogenesis detection of mechanical stimulus involved in sensory perception of sound detection of mechanical stimulus involved in equilibrioception retina morphogenesis in camera-type eye lens induction in camera-type eye epithelial tube branching involved in lung morphogenesis
It has to be done as per old AB suggested Products section.
guarantee_icon

Performance Guarantee

If an Invitrogen™ antibody doesn't perform as described on our website or datasheet,we'll replace the product at no cost to you, or provide you with a credit for a future purchase.*

Learn more
help_icon

We're here to help

Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.

Contact tech support

Your items have has been added!


Host server : magellan-search-5f5c96ff6c-st5hm:80/100.66.129.203:80.
git-commit: 945c3ce0b94337c0c8e9574aed9898c23d0d1109
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.36.1-2025.09.37-1.0