Disclaimer

Clicking the images or links will redirect you to a website hosted by BenchSci that provides third-party scientific content. Neither the content nor the BenchSci technology and processes for selection have been evaluated by us; we are providing them as-is and without warranty of any kind, including for use or application of the Thermo Fisher Scientific products presented.

  • Primary Antibodies ›
  • SERPINB10 Antibodies

Proteintech

SERPINB10 Polyclonal Antibody

2 References
View all (7) SERPINB10 antibodies

{{$productOrderCtrl.translations['antibody.pdp.commerceCard.promotion.promotions']}}

{{$productOrderCtrl.translations['antibody.pdp.commerceCard.promotion.viewpromo']}}

{{$productOrderCtrl.translations['antibody.pdp.commerceCard.promotion.promocode']}}: {{promo.promoCode}} {{promo.promoTitle}} {{promo.promoDescription}}. {{$productOrderCtrl.translations['antibody.pdp.commerceCard.promotion.learnmore']}}

Datasheet
Protocols
Questions & Answers
Datasheet
Protocols
Questions & Answers

Cite SERPINB10 Polyclonal Antibody

  • Antibody Testing Data (11)
SERPINB10 Antibody in Immunocytochemistry (ICC/IF)
Group 53 Created with Sketch.
SERPINB10 Antibody in Immunocytochemistry (ICC/IF)
Group 53 Created with Sketch.

FIGURE: 1 / 11

{{ $ctrl.currentElement.advancedVerification.fullName }}
{{ $ctrl.currentElement.advancedVerification.text }}

SERPINB10 Antibody (21719-1-AP) in ICC/IF

Immunofluorescent analysis of HepG2 cells, using SERPINB10 antibody 21719-1-AP at 1:25 dilution and Rhodamine-labeled goat anti-rabbit IGG (red). {{ $ctrl.currentElement.advancedVerification.fullName }} validation info. View more
Published figure supplied by benchsci-logo
PMID: {{$ctrl.currentElement.benchSciPubmedId}}
View Product

{{$ctrl.videoDesc}}

{{$ctrl.videoLongDesc}}

SERPINB10 Antibody in Immunocytochemistry (ICC/IF)
SERPINB10 Antibody in Immunohistochemistry (Paraffin) (IHC (P))
SERPINB10 Antibody in Immunohistochemistry (Paraffin) (IHC (P))
SERPINB10 Antibody in Immunohistochemistry (Paraffin) (IHC (P))
SERPINB10 Antibody in Immunohistochemistry (Paraffin) (IHC (P))
SERPINB10 Antibody in Immunohistochemistry (Paraffin) (IHC (P))
SERPINB10 Antibody in Western Blot (WB)
SERPINB10 Antibody in Western Blot (WB)
SERPINB10 Antibody in Western Blot (WB)
SERPINB10 Antibody in Western Blot (WB)
SERPINB10 Antibody in Western Blot (WB)
SERPINB10 Polyclonal Antibody

Product Details

21719-1-AP

Applications
Tested Dilution
Publications

Western Blot (WB)

1:500-1:2,000
View 1 publication 1 publication

Immunohistochemistry (IHC)

-
View 1 publication 1 publication

Immunohistochemistry (Paraffin) (IHC (P))

1:20-1:200
-

Immunocytochemistry (ICC/IF)

1:10-1:100
-
Product Specifications

Species Reactivity

Human, Mouse, Rat

Published species

Human, Mouse

Host/Isotype

Rabbit / IgG

Class

Polyclonal

Type

Antibody

Immunogen

SERPINB10 Fusion Protein Ag16372 (1-307 aa encoded by BC096217)
View immunogen

Conjugate

Unconjugated Unconjugated Unconjugated

Form

Liquid

Concentration

0.19 mg/mL

Purification

Antigen affinity chromatography

Storage buffer

PBS, pH 7.3, with 50% glycerol

Contains

0.02% sodium azide

Storage conditions

-20°C

Shipping conditions

Wet ice (domestic); Dry ice (international)

Product Specific Information

Immunogen sequence: MDSLATSIN QFALELSKKL AESAQGKNIF FSSWSISTSL TIVYLGAKGT TAAQMAQVLQ FNRDQGVKCD PESEKKRKME FNLSNSEEIH SDFQTLISEI LKPNDDYLLK TANAIYGEKT YAFHNKYLED MKTYFGAEPQ PVNFVEASDQ IRKDINSWVE RQTEGKIQNL LPDDSVDSTT RMILVNALYF KGIWEHQFLV QNTTEKPFRI NETTSKPVQM MFMKKKLHIF HIEKPKAVGL QLYYKSRDLS LLILLPEDIN GLEQLEKAIT YEKLNEWTSA DMMELYEVQL HLPKFKLEDS YDLKSTLS (1-307 aa encoded by BC096217)

Target Information

Protease inhibitor that may play a role in the regulation of protease activities during hematopoiesis and apoptosis induced by TNF. May regulate protease activities in the cytoplasm and in the nucleus.

For Research Use Only. Not for use in diagnostic procedures. Not for resale without express authorization.

Bioinformatics

Protein Aliases: Bomapin; Peptidase inhibitor 10; PI-10; protease inhibitor 10 (ovalbumin type, bomapin); serine (or cysteine) peptidase inhibitor, clade B (ovalbumin), member 10; serine (or cysteine) peptidase inhibitor, clade B (ovalbumin), member 10, pseudogene; serine (or cysteine) proteinase inhibitor, clade B (ovalbumin), member 10; Serpin B10; serpin peptidase inhibitor, clade B (ovalbumin), member 10; TGF-beta-repressible serine proteinase inhibitor; Transforming growth factor beta repressible serpin; Trespin

View more View less

Gene Aliases: 9830131G07; BB233602; PI-10; PI10; SERPINB10; Serpinb10-ps; Trespin

View more View less

UniProt ID: (Human) P48595, (Rat) Q8K3K4

View more View less

Entrez Gene ID: (Human) 5273, (Mouse) 241197, (Rat) 266775

View more View less

Function(s)
serine-type endopeptidase inhibitor activity peptidase inhibitor activity
Process(es)
positive regulation of defense response to virus by host negative regulation of endopeptidase activity mitophagy in response to mitochondrial depolarization xenophagy negative regulation of peptidase activity negative regulation of proteolysis
It has to be done as per old AB suggested Products section.
guarantee_icon

Performance Guarantee

If an Invitrogen™ antibody doesn't perform as described on our website or datasheet,we'll replace the product at no cost to you, or provide you with a credit for a future purchase.*

Learn more
help_icon

We're here to help

Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.

Contact tech support

Your items have has been added!


Host server : magellan-search-9dd846b7f-s9zn4:80/100.66.128.142:80.
git-commit: 17e8f6c92ed407c129a469238c057b56fdf8abbb
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.36.2-2025.09.39-1.0